25 Jun 2019
Au-Ni-Cl Nanoparticles
This recipe segments nanoparticles of different element from an EDS map.
![front-3 front-3](/software-em-3d-vis/xtra-library/uploads/2019/06/front-3-lg.png)
This recipe segments nanoparticles of different elements from an EDS map. Each element is segmented individually with a given output in the recipe (3 outputs). Then each labeled particles map is used to measure the distribution of the particles. Each distribution is then layed out in the same histogram.
Sample courtesy J. Bursik, Institute of Physics of Materials, Brno