18 Nov 2021
How to Set the Colormap for Ortho Slice and Volume Rendering
This tutorial is an introduction to the basic principle and use of colormaps.
This tutorial demonstrates the basic use of the colormap port of display modules (here an Ortho Slice and a Volume Rendering) in the following ways:
- edit the colormap range,
- use the alpha channel to play on transparency,
- edit the opacity,
- pick a different colormap.
Additional resources: