14 Sep 2021
How to Change the Label Colormap to Display a Label Field
This short video explains how to set an appropriate colormap for label fields with a large number of labels.
A label colormap maps one color value to each label of a label field. By default, Amira-Avizo uses labels.am for rendering label fields, but this colormap contains only 8 colors. Therefore, it is useful to know how to change the label colormap for label fields with more than 8 labels; for example, to labels256.am, as demonstrated in this video.
Additional resources: