24 Jul 2019
Plates in Ti-62222
FIB Serial Section of Ti-62222
The recipe is composed of the following steps:
- Remove curtaining artefacts
- Remove high frequency signal
- Remove the Beta phase and estimate an image of the background
- Perform a background correction
- Binarize the Beta phase and get the Alpha phase