26 Jun 2019
Porosity in a Polymer Divider
The recipe detects pores in a polymer divider (SEM image).
front-6
The recipe detects pores in a polymer divider. Pores are labeled individually to measure area, length, width, equivalent diameter etc. The labeled pores are colored according to their area.