26 Jun 2019
Size distribution of Nb-Cu nanoparticles
The recipe segments the Nb particles and the Cu particles.
![front-8 front-8](/software-em-3d-vis/xtra-library/uploads/2019/06/front-8-lg.png)
The recipe segments the Nb particles and the Cu particles.
The blue channel is extracted and an hysteresis thresholding is performed to segment the Nb particles. The same procedure is applied to the green channel for Cu particles.