Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ATG8H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111517.3 | NP_566283.1 | 4 - 5 | 396 | 108 | |
GenBank mRNA | AB073182.1 | - | 4 - 5 | 294 | 108 | |
AK176187.1 | - | 4 - 5 | 396 | 108 | ||
AK176657.1 | - | 4 - 5 | 396 | 108 | ||
AY085270.1 | - | 4 - 5 | 392 | 108 | ||
BT024566.1 | - | 4 - 5 | 273 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |