Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003050.1 | NP_001003050.1 | 2 - 3 | 125 | 93 | |
XM_005633269.3 | XP_005633326.1 | 2 - 3 | 227 | 93 | ||
XM_005633270.2 | XP_005633327.1 | 2 - 3 | 149 | 93 | ||
GenBank mRNA | M59172.1 | - | 2 - 3 | 125 | 93 | |
M59173.1 | - | 1 - 2 | 69 | 93 |
Gene Symbol: SAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313872.1 | NP_001300801.1 | 2 - 3 | 178 | 93 | |
XM_005633730.3 | XP_005633787.1 | 2 - 3 | 159 | 93 | ||
GenBank mRNA | M59171.1 | - | 1 - 2 | 93 | ||
M59174.1 | - | 1 - 2 | 55 | 93 | ||
M59175.1 | - | 1 - 2 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Inventoried Probe spans exons |