Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rbm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246799.1 | NP_001233728.1 | 1 - 2 | 101 | 60 | |
XM_007645600.1 | XP_007643790.1 | 2 - 3 | 219 | 60 | ||
XM_007645602.1 | XP_007643792.1 | 2 - 3 | 219 | 60 | ||
GenBank mRNA | EU556497.1 | - | 1 - 2 | 101 | 60 | |
JI874849.1 | - | 3 - 4 | 627 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |