Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: rdh8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200788.2 | NP_957082.2 | 1 - 2 | 179 | 91 | |
GenBank mRNA | AY306006.1 | - | 1 - 2 | 177 | 91 | |
BC059640.1 | - | 1 - 2 | 179 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |