Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppp1ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031868.2 | NP_114074.1 | 2 - 3 | 226 | 79 | |
GenBank mRNA | AK007932.1 | - | 2 - 3 | 227 | 79 | |
AK028392.1 | - | 2 - 3 | 216 | 79 | ||
AK090070.1 | - | 2 - 3 | 244 | 79 | ||
AK150344.1 | - | 2 - 3 | 244 | 79 | ||
AK151582.1 | - | 2 - 3 | 244 | 79 | ||
AK152051.1 | - | 2 - 3 | 244 | 79 | ||
AK152585.1 | - | 2 - 3 | 244 | 79 | ||
AK152667.1 | - | 2 - 3 | 222 | 79 | ||
AK153517.1 | - | 2 - 3 | 208 | 79 | ||
AK158681.1 | - | 2 - 3 | 182 | 79 | ||
AK159575.1 | - | 2 - 3 | 234 | 79 | ||
AK167244.1 | - | 2 - 3 | 227 | 79 | ||
AK167538.1 | - | 2 - 3 | 239 | 79 | ||
AK167880.1 | - | 2 - 3 | 228 | 79 | ||
AK167981.1 | - | 2 - 3 | 227 | 79 | ||
AK197966.1 | - | 1 - 2 | 57 | 79 | ||
AK218337.1 | - | 1 - 2 | 57 | 79 | ||
BC014828.1 | - | 2 - 3 | 232 | 4239005 | 79 | |
U25809.1 | - | 2 - 3 | 247 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |