Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Xab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026156.2 | NP_080432.1 | 10 - 11 | 1392 | 67 | |
GenBank mRNA | AK002890.1 | - | 10 - 11 | 1393 | 67 | |
AK132434.1 | - | 10 - 11 | 1406 | 67 | ||
AK145925.1 | - | 10 - 11 | 1394 | 67 | ||
BC021341.1 | - | 10 - 11 | 1390 | 5066146 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |