Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mthfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161798.1 | NP_001155270.1 | 6 - 7 | 1174 | 73 | |
NM_010840.3 | NP_034970.2 | 6 - 7 | 1097 | 73 | ||
NR_027809.1 | 6 - 7 | 1174 | 73 | |||
GenBank mRNA | AK030192.1 | - | 6 - 7 | 1092 | 73 | |
AK150159.1 | - | 6 - 7 | 1175 | 73 | ||
BC051017.1 | - | 6 - 7 | 1076 | 6308248 | 73 | |
BC052466.1 | - | 6 - 7 | 1141 | 6834886 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |