Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Zcchc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035509.1 | NP_001030586.1 | 2 - 3 | 303 | 93 | |
NM_025893.2 | NP_080169.2 | 2 - 3 | 251 | 93 | ||
GenBank mRNA | AK005317.1 | - | 2 - 3 | 229 | 93 | |
BC017627.1 | - | 2 - 3 | 164 | 4505033 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |