Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZWINT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960303.1 | XP_027816104.1 | 7 - 8 | 861 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |