Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pdia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053849.1 | NP_446301.1 | 7 - 8 | 1412 | 67 | |
GenBank mRNA | BC061535.1 | - | 7 - 8 | 1226 | 6922170 | 67 |
M86870.1 | - | 7 - 8 | 1412 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |