Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019209.1 | NP_062082.1 | 3 - 4 | 249 | 60 | |
GenBank mRNA | BC158690.1 | - | 3 - 4 | 249 | 8376316 | 60 |
DY314066.1 | - | 3 - 4 | 249 | 60 | ||
EV767503.1 | - | 2 - 3 | 207 | 60 | ||
FQ216026.1 | - | 3 - 4 | 242 | 60 | ||
FQ216320.1 | - | 3 - 4 | 225 | 60 | ||
FQ218910.1 | - | 3 - 4 | 240 | 60 | ||
FQ230458.1 | - | 3 - 4 | 222 | 60 | ||
FQ233169.1 | - | 3 - 4 | 226 | 60 | ||
FQ234620.1 | - | 3 - 4 | 247 | 60 | ||
FQ234801.1 | - | 3 - 4 | 226 | 60 | ||
FQ235247.1 | - | 3 - 4 | 226 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |