Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dhrs7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271597.1 | NP_001258526.1 | 5 - 6 | 726 | 98 | |
NM_001271598.1 | NP_001258527.1 | 5 - 6 | 726 | 98 | ||
GenBank mRNA | FQ214964.1 | - | 5 - 6 | 706 | 98 | |
FQ215595.1 | - | 5 - 6 | 707 | 98 | ||
FQ216481.1 | - | 5 - 6 | 708 | 98 | ||
FQ224852.1 | - | 5 - 6 | 707 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Rattus norvegicus Probe spans exons |