Search Thermo Fisher Scientific
Axiom myDesign arrays allow flexible access to custom genotyping for small and large studies. They are supported by a world-class design team for fast design and delivery. With Axiom myDesign Genotyping Arrays, you can create fully or semi-customizable genotyping arrays for agrigenomics and human genotyping applications. Based on the universal workflow of the Axiom Genotyping Solution, Axiom myDesign Arrays offer unprecedented scale and flexibility to design the most optimal array for your species of interest. Fully or semi-customized arrays containing 1,500 to 2.6 million markers can be designed and available in as few as 6 weeks after SNP lockdown.
Consortia members interested in developing a new array can request a design consultation by contacting our chip design team at BioinformaticsServices@thermofisher.com.
Axiom myDesign Arrays offer superior scale and flexibility to design the most optimal array for your disease and population studies involving genome-wide association, replication, fine mapping, and candidate gene studies. Include markers of your choosing or select markers from the Axiom Genomic Database, which contains over 10 million high-performance genotype-tested (biologically and technically validated) annotated common and rare variants. Proprietary imputation-based SNP selection algorithms can be used to design population-optimized GWAS marker modules that deliver high genomic coverage and accuracy.
Flexible optimized solutions provide breeders and researchers with powerful and affordable genotyping tools to identify, validate, and routinely screen complex genetic traits for leveraging the highest economic value of the herd or plant. Our portfolio of robust, highly reproducible, and scalable products facilitates marker-assisted selection, genome-wide association studies, quantitative trait loci analyses, parentage, and traceability, all of which fast track your genomic selection programs. Customize your genotyping array or panel with us for any high throughput industrial application with confidence that all your markers of interest can be targeted and manufactured on the custom array or panel.
Axiom myDesign Custom Arrays feature:
Axiom myDesign Arrays are available in formats suitable for the appropriate number of markers:
Axiom myDesign Genotyping Arrays include:
Cat. No. | Product | |
---|---|---|
000870 | Axiom 384HT myDesign Custom Array |
|
000928 | Axiom Mini 96 myDesign Custom Array, 1.5K - 50K |
|
000841 | Axiom myDesign TG Array, 1K-90K |
|
000799 000919 (1x24) | Axiom myDesign TG Array, 90K-200K |
|
000780 000920 (1x24) | Axiom myDesign GW Array Plate, 200K-675K |
|
000786 | Axiom myDesign GW Array Plate, 675K-1.3M |
|
000794 | Axiom myDesign GW Array Plate, 1.3M-2.0M |
|
000787 | Axiom myDesign GW Array Plate, 2.0M-2.6M |
|
901758 | Axiom 2.0 Reagent Kit | Includes all reagents (except isopropanol) to process 96 reactions/plate |
902798 | Axiom 2.0 Reagent Kit 4x24 Reactions | Includes all reagents (except isopropanol) to process 4 x 24 array plates |
902245 | Axiom 2.0 384HT Reagent Kit | Includes all reagents to process one Axiom 384HT Array Plate |
901606 | Axiom GeneTitan Consumables Kit | Contains all GeneTitan consumables required to process one Axiom Array Plate |
902234 | Axiom 384HT GeneTitan Consumables Kit | Includes consumables required to process one Axiom 384-Array Plate on the GeneTitan Instrument |
902289 | Axiom 384HT Consumables Kit for QC | Includes all consumables required for QC of ten Axiom 384-Array Plates |
902288 | Axiom 384HT Consumables Kit for Biomek FXp | Includes consumables required for target prep on the Biomek FXp for five Axiom 384-Array Plates |
903013 | Axiom 2.0 Assay Mini 96 Reagent Kit | Includes reagents to process two Axiom Mini 96 format array plates |
903014 | Axiom Mini 96 Consumable Kit for QC | Includes all consumables required for QC of ten Mini 96 array plates |
903015 | Axiom 2.0 Assay Mini 96 Training Kit | Includes reagents, consumables and DNA required for training |
902986 | Axiom 2.0 Assay Mini 96 Manual Target Preparation Consumables Kit | Includes consumables for manual target preparation of four Mini 96 array format plates |
902629 | Axiom 384HT High Volume Consumable Kit | Includes consumables for processing five Mini 96 arrays on the GeneTitan MC instrument |