Organism TypeMouseHybridoma cells derived from fusion of Sp2/0-Ag14 cells and BALB/c mouse splenocytes.
TissuesSpleen
PhenotypeSuspension
Primaryno
ApplicationCell Culture / Growth Conditions
Products
Catalog # Name Size Price (USD) Qty
11965092 DMEM, high glucose, 500 mL Each
34.65
Online Exclusive
37.14
Save 2.49 (7%)
12045084 Hybridoma-SFM, 500 mL Each
69.65
Online Exclusive
77.00
Save 7.35 (10%)
26140079 Fetal Bovine Serum, qualified, United States, 500 mL Each -
Every step of the way, a wide range of cell health products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.