c7c8 - Key/Specs/Resources/Appl/Tech/Doc/Contact
The rapid innovations in advanced 2.5D and 3D packaging, complex interconnect schemes, and higher-performance power devices are creating unprecedented failure localization and analysis challenges. Defective semiconductor devices often show a variation in the local power dissipation, leading to local temperature increases. The Thermo Scientific ELITE System utilizes lock-in IR thermography (LIT) to accurately and efficiently locate these areas of interest and provide non-destructive 3D device insights. The ELITE System’s optics and InSb camera are specifically designed to achieve high localization resolution and sensitivity and solve the most difficult analytical challenges.
LIT, a form of dynamic IR thermography, provides maximum signal-to-noise ratio, increased sensitivity, and higher feature resolution compared to steady-state thermography. It is the ideal solution for the localization and analysis of line shorts, ESD defects, oxide damage, defective transistors, diodes, and device latch-ups. An optional laser scanning microscope enables high-resolution OBIRCH to complement the ELITE System’s thermal analysis capabilities.
The ELITE System is also available in the VX configuration, which provides all the features required for localizing defects in advanced power devices based on Si, SiC, or GaN technologies.
Video Player is loading.
Current Time 0:00
Duration 5:42
Loaded: 0.00%
Stream Type LIVE
Remaining Time 5:42
1x
    • Chapters
    • descriptions off, selected
    • en (Main), selected
    On-demand Spark Webinar: Semiconductor Packaging Challenges
    Watch on demand
    Key Features
    • Very high sensitivity, InSb camera, and thermal emission optics enable nondestructive, through-package, and stacked die analysis
    • Real-time lock-in measurement
    • Contactless absolute temperature mapping
    • Optical beam-induced resistance change (OBIRCH) option
    • High-voltage power device analysis option (VX)
      West Coast Failure Analysis Seminar: Ask the Expert
      Register for webinar
    Specifications
    Style Sheet for Products Table Specifications
    Lateral resolution
    • Down to 1 μm
    Depth resolution
    • Down to 20 μm
    Defect types
    • Wide range of shorts (2 mΩ to 2 GΩ), leakage (power dissipation as low as 1 μW), resistive opens
    Sample types
    • Board assemblies, modules, packages, full wafers, wafer coupons, die
    FOV
    • Max 200 mm x 160 mm; min 0.62 mm x 0.51 mm
    DUT stimulation
    • Internal DC power supply; ATE, CA bus, boundary scan tester, system level tester
    • Up to 10 kV capable (requires VX option)
    Time to results
    • Minutes to seconds, depending on applied power and sample
    *Performance may vary depending on sample and specific setup.
    Style Sheet for Komodo Tabs
    Resources
    The ability to locate a thermal source in 3D makes the ELITE system well-suited for stacked-die analysis.
    The ability to locate a thermal source in 3D makes the ELITE system well-suited for stacked-die analysis.
    Sensitivity varies with power level
    Sensitivity varies with power level.
    The ability to locate a thermal source in 3D makes the ELITE system well-suited for stacked-die analysis.
    The ability to locate a thermal source in 3D makes the ELITE system well-suited for stacked-die analysis.
    Sensitivity varies with power level
    Sensitivity varies with power level.
    Applications
    Semiconductor Device Packaging
    Semiconductor Advanced Packaging
    Performance, power efficiency, area, and cost are driving packaging innovations. Learn how workflows provide fast, precise, and accurate time-to-data.
    Semiconductor-RD_1920x1080
    Semiconductor research and development
    Innovation starts with research and development. Learn more about solutions to help you understand innovative structures and materials at the atomic level.
    Semi-Metrology_1920x1080
    Semiconductor metrology
    Manufacturing today’s complex semiconductors requires exact process controls. Learn more about advanced metrology and analysis solutions to accelerate yield learnings.
    Semiconductor Failure Analysis
    Semiconductor Failure Analysis
    Complex semiconductor device structures result in more places for defects to hide. Learn more about failure analysis solutions to isolate, analyze, and repair defects.
    esd_thumb_274x180_144dpi
    ESD Semiconductor Qualification
    Every electrostatic discharge (ESD) control plan is required to identify devices that are sensitive to ESD. We offer a complete suite of test systems to help with your device qualification requirements.
    Semiconductor power devices
    Semiconductor power devices
    Novel architectures and materials pose new challenges. Learn how to pinpoint faults and characterize materials, structures, and interfaces.
    Display Module Failure Analysis
    Semiconductor display technology
    Display technologies are evolving to improve display quality and light conversion efficiency. Learn how metrology, failure analysis, and characterization solutions provide insights.
    Semi-Materials-Character_1920x1080
    Semiconductor materials characterization
    Many factors impact yield, performance, and reliability. Learn more about solutions to characterize physical, structural, and chemical properties.
    Techniques
    Thermal Fault Isolation
    Uneven distribution of local power dissipation can cause large, localized increases in temperature, leading to device failure. We offer unique solutions for thermal fault isolation with high-sensitivity lock-in infrared thermography (LIT).
    Learn more ›
    Thermal Fault Isolation
    Uneven distribution of local power dissipation can cause large, localized increases in temperature, leading to device failure. We offer unique solutions for thermal fault isolation with high-sensitivity lock-in infrared thermography (LIT).
    Learn more ›
    Style Sheet to change H2 style to p with em-h2-header class
    Contact us
    Electron microscopy services for
    semiconductors
    To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
    Learn more ›
    Style Sheet for Support and Service footer
    Style Sheet for Fonts
    Style Sheet for Cards