Sterilin Universal Containers
Protect your valuable samples
Ideal for applications where safety is a concern, as when handling valuable or potentially hazardous samples, these leakproof containers—designed for unrivalled ease of handling–minimize sample loss and suit the needs of most laboratories.
Facilitate sample collection and transportation to the lab in a single container using Thermo Scientific™ Sterilin™ Dippas™ to reduce risk of cross-contamination.
For small-volume samples, choose Thermo Scientific™ Sterilin™ disposable glass containers which are made from soda glass with leakfree polypropylene caps.
Use Thermo Scientific™ Sterilin™ disposable polypropylene containers for a wide range of sampling applications. 
Perform sterility and endotoxin tests with Thermo Scientific™ Sterilin™ non-pyrogenic polystyrene containers.
Use Thermo Scientific™ Sterilin™ triple bagged polystyrene containers in hospital rooms and other such sterile areas.
Style Sheet for Global Design System