Structure guided development of cell and gene therapies
Advancements in cell and gene therapy are transforming the landscape of how we deal with disease. Cell therapy harnesses the power of cells to treat diseases ranging from cancer to genetic disorders. Gene therapy involves the introduction, removal, or modification of genetic material within a patient’s body to treat or prevent diseases. For both of these approaches, from rational design for CAR T-cell receptor to gene delivery vehicle characterization, cryo-electron microscopy (cryo-EM) is continuing to play an important role.
Rational design for CAR T-cell receptor
CAR T cell therapy represents a popular approach in the field of cell therapies, harnessing the power of patients' own T cells to identify and eradicate cancer cells. At the core of CAR T-cell therapy lies the Chimeric Antigen Receptor (CAR) receptor, a modular receptor assembled from components derived from immune elements. The antigen-binding domain on the CAR receptor recognizes and binds specific antigens present on the surface of cancer cells, initiating downstream signaling that triggers the destruction of cancer cells. To achieve optimal affinity tuning and specificity, a detailed understanding of the high-resolution 3D protein structure of CAR receptors is important.
Schematic of the CAR receptor in complex with CD19 antigen displayed on a tumor cell
CAR (chimeric antigen receptor) on a T cell in complex with CD19 antigen displayed on a tumor cell.
Cryo-EM enables the study of 3D structure of protein complexes at near-atomic resolution. The 3D structure of CAR receptor in complex with cancer antigens, provides insights into their atomic-level interactions and conformational changes. Such details enable researchers to precisely engineer CAR receptors with suitable affinity and specificity, maximizing their therapeutic potential. A study published in Science Immunology showed the application of cryo-EM to solve the molecular structure of CAR receptor bound to CD19, a target antigen for the blood cancers. This information then was used to design several CAR receptors with distinct binding affinity towards CD19 and to generate CAR T cells with unique cancer cell killing capabilities.
Illustration of gene therapy process
Gene therapy involves the delivery of gene editing machinery or a replacement gene to a desired cell/tissue with the help of a suitable delivery vehicle.
Discovery and engineering of gene editor enzymes
Genome editing has become a rapidly growing field for the treatment and prevention of various diseases, as this technology allows for the precise manipulation of a specific genetic sequence inside an organism. Several advanced techniques have emerged, with CRISPR-Cas9 being a prominent example. Recent advancements in cryo-EM technology enables scientists to not only discover new gene editing enzymes but also to perform structure-guided engineering to improve gene editing properties of these enzymes.
There are generally 3 types of tools used:
  • CRISPR-Cas9 - Comprises clustered regularly interspaced short palindromic repeats (CRISPR) and CRISPR associated protein 9 (Cas9). The CRISPR-Cas9 system permits targeted gene cleavage and gene editing in a variety of cells.
  • Zinc finger nucleases – These were among the first programmable nucleases designed for precise genome editing.
  • TALENs or Tal effectors – Used for highly precise and efficient gene editing in live cells
Discovery and elucidation of mechanism of action of novel gene editors
Cryo-EM structure of SpCas9-ABE8e complex
Cryo-EM structures reveal gene editors in their native state and elucidates the distinct mechanisms of genome recognition and editing. Structure of SpCas9-ABE8e complex from Lapinaite A et al, Science, 2020
Structure-guided engineering of gene editors
Cryo-EM structure of Cas9 18-20 MM
Cryo-EM structures are used to improve the size, accuracy, speed, and other biochemical properties of existing enzymes or lead to design completely new enzymes. Structure of Cas9 18-20 MM from Bravo J et al, Nature, 2022
To realize the full therapeutic potential of gene editing systems, it is critical to reduce off-target gene editing, ensure precise editing at the desired target, and developing stable and targetable delivery vehicles. Understanding the structural basis of how these enzymes recognize the target and perform gene editing function will open up new avenues for introducing rational changes to gene editing enzymes to improve their efficiency while avoiding off-targets effects. The use of single particle cryo-EM has been increasingly used to determine the structures of gene editing enzymes. A recent paper by Nakagawa et. al2., used cryo-EM to gain structural insights on CRISPR-Cas12 effectors, a later addition to the CRISPR family but ones that are gaining traction as powerful tool for non-mammalian applications and diagnostics.
Gene delivery vehicle design and characterization
Success of gene therapy is directly correlated with the efficiency of the gene delivery vehicles. These delivery vehicles ensure the targeted delivery of gene editors/gene replacements to specific cells or tissue within the body. Gene delivery vehicles can be generally classified into two categories:
  • Viral vectors like Adeno-associated viruses (AAV) and Lentiviral-based vectors (LVV)
  • Non-viral vectors like Liposomes, Lipid nanoparticles (LNPs) and Lipoplexes  
Cryo-EM workflow for capsid characterization
Image adapted from Hsu HL et al, Nature Communication, 2020
Structure guided capsid engineering
Capsid engineering involves the deliberate modification of capsid of known AAV serotypes or the engineering of a new type of viral capsid. The goal for capsid engineering is to generate a vector system that can go undetected by the immune system (reduced immunogenicity), can escape neutralization by pre-existing antibodies and is able to target a specific cell or tissue type (tropism). Single particle cryo-EM offers insights into the molecular structure of novel viral capsids and the molecular basis of viral capsid binding to the cell surface receptors or neutralizing antibodies. Once the region of interest is identified, precise structural modification can be made to improve on the desired capsid properties.
Cryo-EM assisted capsid engineering
Discovery and characterization of novel viral vectors
High-resolution capsid structure provides a platform for capsid engineering. Structural details can also be used to identify new features for novel viral vector identification, regulatory approval as well as for Intellectual property (IP) protection. Hsu et.al3, published research detailing a novel capsid which shared sequence similarity with AAV2, one of three serotypes approved for commercial use in patients. Their 2.5 Å resolution structure identified critical amino acid residues that contributed to the characteristics of their novel vector that they obtained from clinical samples. 
Density map of AAVv66
Density map of new AAV capsid. Figure from Hsu et. al. 2020 under a creative commons license.
Viral capsid-receptor interaction
Mapping of receptor interaction sites help facilitate AAV engineering with enhanced or unique targetability. Furthermore, capturing these small differences on how different AAV serotypes interact with cell surface receptors and utilizing them to train artificial intelligence/machine learning algorithms will help generate diverse AAV capsids with unique specificity and targetability.
Zhang R et al used single particle cryo-EM to understand the molecular basis cell surface receptor binding of three AAV serotypes (AAV1, AAV2 and AAV5).
Cryo-EM analysis of AAVR positions on AAV capsids
Image adapted from Zhang R et al, Nature Communications, 2019, illustrating the use of cryo-EM to understand receptor binding of three AAV serotypes.
Capsid Immunogenicity optimization
Cryo-EM structures provide insights into AAV neutralization mechanisms and to rational design of escape variants with minimal disruption to cell tropism and gene expression. Smith JK and Agbandje-McKenna M, used cryo-EM to identify neutralizing antibody (Nab) hotspots on the vector surface and combined this information to perform site directed mutagenesis to eliminate these hotspots to create viral vectors with the ability to avoid preexisting immunity against gene delivery vectors.
Cryo-EM analysis for rational design of virtual vectors
Image adapted from Smith JK and Agbandje-McKenna M, PLOS Pathogens, 2018, illustrating the use of cryo-EM to visualize areas of antibody binding to improve the efficacy of viral vectors for gene therapy.
Viral vector characterization using cryo-EM image analysis
It is well established that an imbalance in the ratio of genome containing AAV particles to the empty AAV particles in a gene therapy formulation affects the therapeutic efficacy. Furthermore, it is important to characterize that capsid engineering efforts did not negatively impact viral capsid properties such as capsid stability. Production process and storage conditions can also negatively affect the stability, integrity, and infectivity of virus particles. Therefore, comprehensive viral vector characterization is essential to ensures efficacy and safety of gene therapy products.
Determination of empty-full AAV capsid ratio
The ratio of empty to full viral particles is a critical factor that affects the overall efficacy of AAV based gene therapy. An imbalanced ratio can reduce the therapeutic payload delivered to target cells, compromising the desired therapeutic effect. Accurate characterization of the empty-to-full ratio allows researchers and developers to optimize vector production and ensure maximum efficiency in delivering the therapeutic cargo.
Regulatory agencies around the world such as US-FDA and EU-EMA are asking to report accurate quantification of the ratio of empty vs. full particle in gene therapy formulations.
Cryo-EM analysis of AAV viral particles
Analysis of empty to full ratio of viral particles for AAV based gene therapy File: empty to full sysmatic_2048
Revealing particle morphology
The structural integrity and morphology of viral vectors play a vital role in their ability to effectively deliver therapeutic genes. 
Aggregated and broken viral particles are considered as impurities and their accurate estimation is required by regulatory agencies. Such information can also be utilized in the process optimization stage to optimize the viral vector production process.
Cryo-EM analysis for the morphology of viral vectors
Decoding surface features
The surface properties of viral vectors determine their interactions with target cells, host immune responses, and potential risks associated with off-target effects. Cryo-EM provides the unique capability to capture detailed images of the viral vector's surface structure, allowing for the identification of critical features such as smooth capsid verse capsid with a spiky surface in case of Lentivirus based viral vectors. This high-resolution information enables scientists to fine-tune vector design and enhance their specificity, reducing potential adverse effects and improving overall therapeutic efficacy.
High resolution imaging of viral vectors
Cryo-EM projection images of Env− HIV-1 mature VLPs and SIV virions from cryo-electron tomography. Image adapted from Ni T et al., Structure Biology, 2021 and Zanetti G et al., PLOS Pathogens, 2006.
Recommended further reading:
  1. He C, Mansilla-Soto J, Khanra N, Hamieh M, Bustos V, Paquette AJ, Garcia Angus A, Shore DM, Rice WJ, Khelashvili G, Sadelain M, Meyerson JR. CD19 CAR antigen engagement mechanisms and affinity tuning. Sci Immunol. 2023
  2. Nakagawa, R., Hirano, H., Omura, S.N. et al. Cryo-EM structure of the transposon-associated TnpB enzyme. Nature 616, 390–397 (2023).
  3. Hsu, HL., Brown, A., Loveland, A.B. et al. Structural characterization of a novel human adeno-associated virus capsid with neurotropic properties. Nat Commun 11, 3279 (2020).
  4. Nancy L. Meyer and Michael S. Chapman. Adeno-associated virus (AAV) cell entry: structural insights. Trends in Microbiology (2021).
  5. Scott M. Stagg, Craig Yoshioka, Omar Davulcu, and Michael S. Chapman Chemical Reviews 2022 122 (17), 14018-14054
Cryo-EM solutions
Our full cryo-TEM portfolio features state-of-the-art technology with a range of automation features designed to extend accessibility, reduce the need for user intervention, and enable easy organization, viewing, and sharing data.
Glacios 2 Cryo-TEM
A complete solution for high-resolution structure determination with unparalleled automation and ease-of-use.
Krios 5 Cryo-TEM
Atomic resolution cryo-EM instrument with enhanced productivity and compact design.
Tundra Cryo-TEM
Dedicated structural analysis solution designed to bring cryo-EM to your laboratory.
Talos L120C
Versatile TEM and STEM microscope for 2D and 3D visualization of beam sensitive samples and materials using high contrast
Resources
Cryo-EM structure of AAV8 and epitope mapping of CaptureSelect AAVX
Leveraging cryo-electron microscopy for innovative vaccine development