Leveraging cryo-EM for antibody discovery and epitope mapping
Large-molecule antibody biologics have revolutionized drug discovery owing to their superior target specificity and amenability to versatile engineering.  As of 2023, more than 160 monoclonal antibody therapies have been approved by at least one regulatory agency in the world for a wide range of indications. The approved antibody therapies include canonical antibodies, antibody-drug conjugates (ADCs), bispecific antibodies, antibody fragments, immunoconjugates and Fc-Fusion proteins. Cryo electron microscopy (cryo-EM) has emerged as a crucial tool in accelerating the entire process, starting from protein target selection to the development of therapeutic antibodies.
Epitope mapping with cryo-EM
The goal of an antibody discovery pipeline is to develop a stable antibody drug with required specificity, selectivity and affinity that can be patented and/or progressed to clinic.  Epitope mapping is an established process used in multiple stages of the antibody discovery workflow that provides information about the antigen-antibody interaction and helps to understand the mode of action. Detailed epitope information is important to strengthen intellectual property (IP) claims.
Epitopes can be classified into two groups - Linear epitopes, and conformational epitopes. Although, there are several technologies available to characterize the epitope-paratope interaction, cryo-EM offers several advantages over these technologies:
  • Accurate epitope mapping of the epitope and the paratope at a single amino acid level, even of challenging and/or flexible targets.
  • Determine the spatial arrangement of discrete and overlapping epitopes (both linear and conformational) on the large antigen surface.
  • The technique does not require labeling or immobilization and is free from crystallization artifacts.
  • Provides structural insights early on, driving faster decision-making which saves both cost and time.
Antigens can have multiple epitopes that can be revealed by cryo-EM
More than one epitope can be found on an antigen.
Cryo-EM is a rapid, robust, versatile, and widely available method that supports the antibody discovery and development process.  The role of cryo-EM based structure biology can be grouped into three categories based on the antibody function.
Epitope mapping to develop therapeutic antibodies
Epitope mapping using cryo-EM offers high-resolution information at the level of a single amino acid. Such detailed information can help elucidate the mode of action of antibody therapeutics. Having a structural basis of the antigen-antibody interaction can lead to the design of modular antibody formats like bispecific antibodies and ADCs. 
High-throughput cryo-EM epitope mapping of SARS-CoV-2 Spike Protein antibodies
Immunogenicity profile and epitope mapping to develop effective antibodies
Cryo-EM Polyclonal Epitope Mapping (Cryo-EMPEM) is growing in popularity to study the immune response against biologics such as antibodies and vaccines. This workflow allows you to study the polyclonal antibody response against an immunogen/vaccine candidate. Antigen bound to these antibodies can directly be isolated from the serum and subjected to high-resolution cryo-EM analysis. Detailed structural information allows the mapping of different epitopes (antigenic part) on the immunogen (vaccine/therapeutic antibody). These epitopes can then be engineered to enhance the immunogenicity for vaccine design and reduce the immunogenicity for antibody therapeutics and gene therapy viral vectors.
CryoEMPEM analysis of immune complexes generated using polyclonal Fabs
High resolution CryoEMPEM mapping at the amino acid level of neutralizing antibodies binding to different epitopes. Image adapted from Antanasijevic A et al 2021.
Epitope mapping to develop antibodies as use as a tool or reagent
Due to their remarkable selectivity, antibodies are routinely used to develop highly specific diagnostic tests, and affinity resins are used in bioprocessing and downstream purification. More recently, antibody-derived fragments are also being used as a tool to study other therapeutically relevant target proteins such as GPCR and Ion channels. Some of these tools are nanobody, megabody, legobody and NabFab.
Cryo-EM structure of PepT2
Cryo-EM density of solute transporter PepT2-nanobody complex. Image adapted from Parker J et al 2021.
Cryo-EM structure of RBD domain of SARS-CoV-2 spike protein and Legobody
Cryo-EM structure of a complex of RBD domain of the SARS-CoV-2 spike protein and Legobody. Image adapted from We X and Rapoport T, 2021
Antibody discovery workflow
The typical antibody discovery workflow is a long and tedious process that involves multiple steps to develop a successful antibody candidate. Of all the antibody therapeutics that enter phase 1 clinical trials, only 22% go on to receive regulatory approval. Structural information afforded by cryo-EM early in the discovery and development process is an important accelerator of this process.  This information can drive structural modification to improve the developability and alter antibody properties in informed manner. Later in the pipeline, cryo-EM can be used to structurally characterize immunogenicity of candidate antibody in preclinical models and clinical trial stages. Below, we highlight where structural information can drive faster and better-informed decision making. 
Antibody discovery workflow and the information that can be obtained by cryo-EM to accelerate the process.
Hit to lead in therapeutic antibody development
When selecting lead antibody candidates from a large hit dataset, structural information provides detailed epitope binding information and informs on the developability aspect of the antibody molecule. Cryo-EM can help identify the most promising lead antibodies for lead optimization.
Structure elucidation by cryo-EM provides:
  • Resolution of the spatial arrangement of discrete and overlapping epitopes (both linear and conformational) on the large antigen surface
  • Accurate mapping of both the epitope and the paratope at single amino acid level thus providing information on mode of action.
  • Quick turnaround of epitopes at moderate to high-resolution with well resolved antigen-antibody interfaces.
Antigens can bind to antibodies in a linear or a conformational matter.
When AI/ML approaches are applied to generate lead antibody candidates, cryo-EM generated structural data will not only help validate those candidates but also provide rich data to train the existing algorithms and drive downstream optimization of identified lead candidates.
Illustration of AI/ML approach to generate lead antibody candidates
Cryo-EM Solutions
Our full cryo-TEM portfolio features state-of-the-art technology with a range of automation features designed to extend accessibility, reduce the need for user intervention, and enable easy organization, viewing, and sharing data.
Glacios 2 Cryo-TEM
A complete solution for high-resolution structure determination with unparalleled automation and ease-of-use.
Krios 5 Cryo-TEM
Atomic resolution cryo-EM instrument with enhanced productivity and compact design.
Tundra Cryo-TEM
Dedicated structural analysis solution designed to bring cryo-EM to your laboratory.
Talos L120C
Versatile TEM and STEM microscope for 2D and 3D visualization of beam sensitive samples and materials using high contrast
Resources
Cryo-electron microscopy is revolutionizing rational drug discovery pipelines
Rational design leveraging routine, high resolution protein structure determination is driving the discovery and development of diverse biologic and small molecule therapies. Cryo-EM delivers rapid epitope mapping on the atomic scale for antibody therapeutics and immune response profiling, supports elucidation of mechanism of action and is also enabling more therapeutic targets than ever before for Structure-Based Drug Design. Whether to modulate binding affinities or to optimize drug stability, all of these questions can be answered in just one day of data collection.
Watch the webinar to hear experts from Sanofi to discuss:
  • An overview of how to quickly go from protein to structure
  • Key examples of multi-specific drugs and structural insights of a CEACAM5-targeting antibody drug conjugate (ADC)
  • How the adoption of Cryo-EM has significantly increased the number of the targets that can be identified for clinical trials in both biopharma and biotech
Cryo-EM structure of AAV8 and epitope mapping of CaptureSelect AAVX
High-throughput cryo-EM epitope mapping of SARS-CoV-2 spike protein antibodies using EPU Multigrid