Mass Spec Software Header
One CDS for your chromatography and routine mass spectroscopy instruments
Our chromatography and mass spectroscopy instruments are intuitive and innovative — ensuring simple operation while achieving better separations and higher sensitivity than ever before.
Thermo Scientific Chromeleon CDS is built on the same principles — simplifying your workflow and giving you superior insights into your data, delivering ease of use, comprehensive instrument control, automated data analysis and flexible reporting.
Learn how Chromeleon CDS supports quantitative mass spectrometry workflows for all separation techniques and MS variants.
Designed for mass spectrometry
Chromeleon 7.2 Chromatography Data System is the first CDS to support mass spectrometry (MS) instrument control and data processing with all main front-end separation techniques (GC, IC, LC) in an enterprise (client/server) environment. This replies to the rapidly growing demand for using mass spectrometric instruments in quantitative analysis for their higher sensitivity and richer detection information. Chromeleon software delivers full control for many Thermo Scientific MS systems, including tuning and instrument calibration.
Mass Spectrometry Instruments
Chromeleon CDS also contains the necessary MS-specific data views, data processing, and reporting capabilities to streamline your chromatography and MS quantitation workflows in one application.
chromatography studio display
Chromatography Studio display of the Total Ion Chromatogram (TIC) and Mass Spectra plots.
By extending Chromeleon CDS beyond chromatography into MS, it is now possible to streamline chromatography and MS quantitation workflows within a single software package and use your MS like any other routine detector.
Mass spectrometry software features
eBook
Compliance-Ready Mass Spectrometry for Bio/Pharmaceutical Analysis and Development eBook
Examine real-use cases of how a networked Chromatography Data System is the best choice for data acquisition and processing for quantitative LC-MS analyses and for ensuring business efficiency, regulatory compliance and data integrity.
Case study
Boosting Productivity of Monoclonal Antibody Development using Chromeleon CDS
Single software platform streamlines operations and supports powerful mass spectrometry (MS) workflows Symphogen is an antibody oncology-focused company, pioneering discovery and early development of highly differentiated monoclonal antibody (mAb) therapeutics.
Case study
Single-vendor LC-MS and CDS system delivers benefits to Fondazione Edmund Mach Chemistry
“Chromeleon 7.2 CDS is a very comprehensive and simple to use software. In a few steps, you can integrate, identify and quantify the interest compounds and create reports, charts and graphs reducing data processing time.”
Technical note
Automated component identification and confirmation based on comparison to a reference mass spectrum
This technical note demonstrates the use of reference mass spectra to increase the confidence in identification of the components and minimize the possibility of false positive identifications when screening for compounds based on full scan spectral library matching.
Featured videos
Chromeleon Combines LC, GC, MS Instrument Control as well as Method and Data Management
Chromeleon Combines LC, GC, MS Instrument Control as well as Method and Data Management
Video Player is loading.
Current Time 0:00
Duration 1:33
Loaded: 0.00%
Stream Type LIVE
Remaining Time 1:33
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextChromeleon Combines LC, GC, MS Instrument Control as well as Method and Data Management
        Chromeleon Combines LC, GC, MS Instrument Control as well as Method and Data Management
      2. Now Playing
        Up NextVideo 63 - Chromeleon CDS: LC-MS Instrument Control
        Seamlessly integrated control of LC-MS via intuitive ePanels
      3. Now Playing
        Up NextChromeleon 7.2 CDS: GC-MS Instrument Control
        Customizable ePanels provide a consistent look and feel to your instruments, now also for GC-MS instruments. The most common controls are available via the home panel tab, while more advanced settings can be accessed from module-specific ePanels.
      4. Now Playing
        Up NextChromeleon 7.2 CDS: Mass Spectral Plot
        The interactive Mass Spectral Plot provides fast access to your mass spectra. Easily show and compare spectra from selected peaks, any time point in the chromatogram, reference spectra, library matches or average spectra.
    Mass spectrometry software topics
    Orbitrap Exploris 480 Vanquish
    LC-MS software
    Chromeleon CDS simplifies your routine quantitative mass spectrometry data acquisition and analysis. It provides full control of Thermo Fisher single/triple quadrupole and high resolution accurate mass (HR/AM) systems using native control drivers.
    MS-specific views including TIC, XIC, Mass Spectra, quantitation channel, and MS component traces streamline data processing with parameters able to be set per component using MS-specific detection algorithms.
    Here is a recent customer interview discussing their use of Chromeleon CDS, their experience of using it with their HR/AM LC-MS system and the benefits they see to having one software for control and processing:
    Discover how Chromeleon CDS supports LC-MS in a fully automated interpretation workflow for finding targets in chemical reactions in this on demand webinar and case study.
    For more information
    isq-ec-single-quad-ms-230x195
    Single quadrupole MS software
    Chromeleon CDS also includes embedded control of the Thermo Scientific ISQ EC and EM single quadrupole mass spectrometers for LC and IC-MS allowing you to focus on successfully using the system with integrated tools to help you quickly learn the MS system and perform analyses.
    hdi-isq-7000-trace-1310-left-270x195
    GC-MS software
    All the advantages of Chromeleon CDS are now available to GC-MS users, bringing an established and widely-accepted CDS with many unique data processing and reporting features to what has traditionally been a workstation-based technique.
    Chromeleon software offers data organized at the component level with advanced tools for fast and easy visualization of these large amounts of data. Reporting has been enhanced to make it easy to report components based on their specific channel, and NIST format library searching supports component identification and direct import of compound data, such as quantitation and confirmation ions, into the component table.
    Thermo Scientific MS systems can be used with all of Chromeleon software’s productivity features including eWorkflow procedures. See how Broughton Laboratories uses Chromeleon CDS to ensure high-quality results and streamline operations for their GC-MS system.
    For more information
    IC-MS software
    IC-MS software
    With Chromeleon CDS you can increase the power of ion chromatography (IC) with MS detection.
    MS detection enables you to ensure analytical confidence and dramatically improve the detection capability of your Thermo Scientific Dionex Integrion or ICS-6000 HPIC system. IC-MS maximizes the ability to detect and quantify unexpected co-elution of components and to confirm trace components.
    Mass spectrometry software related products
    xps-software-icon-150x150
    Easy-to-Use MS interface: walk up and run
    Thermo Scientific Chromeleon XPS software is an Open Access tool enabling users to simply walk up and use a very straightforward interface to exploit the full power of Chromeleon CDS, without actually using the CDS software itself.
    The Chromeleon XPS interface provides a streamlined and easy-to-use interface for Chromeleon software, while maintaining full capabilities of the CDS, like instrument control, data processing, reporting, and compliance tools.
    headset-icon-150x150
    Contact us
    Want to learn more about how Chromeleon software can help you drive your entire laboratory process, ensure data integrity and compliance and increase productivity? Contact us to now unlock the value of your data.
    Request contact
    Style Sheet for Global Design System
    Style Sheet for Komodo Tabs
    CMD SchemaApp code