Ion Chromatography Mass Spectrometry (IC-MS)
Increase the power of IC with MS detection
Utilize mass detection when you need to ensure analytical confidence and dramatically improve the detection capability of your Thermo Scientific Dionex Integrion or ICS-6000 HPIC system. Ion chromatography with mass spectrometry (IC-MS) maximizes the ability to detect and quantify unexpected co-elutions of components and to confirm trace components.
Browse our complete portfolio below, or let one of our experts help you tailor a selection specific to your application and workflow needs.
Contact us
IC-MS systems
Coupling MS with your IC can give you more information from every sample:
  • Higher sensitivity
  • Accurate quantitation
  • Peak confirmation and evaluation of chromatographic peak purity
  • Increased efficiency
Single Quadrupole IC-MS
Easily master MS for your IC applications.
Triple Quadrupole IC-MS
Need sensitive, fast, and robust quantitation of a variety of molecule types? For complex samples, the high sensitivity and selectivity of MS/MS using selected reaction monitoring (SRM) provides the most precise and accurate quantitation using IC-MS.
Orbitrap IC-MS
Thermo Scientific Orbitrap mass spectrometers generate high-resolution and accurate-mass data, thereby enhancing sensitivity in trace analyte detection, characterization, and quantitation. That's why they are used in research applications—in particular, life science applications such as metabolomics.
Get a superior combination of scan speed, high resolution, mass accuracy, spectral quality and sensitivity for untargeted or targeted screening and a broad range of qualitative and quantitative applications in environmental, food safety and metabolomics.
Use this system for IC-MS in life science metabolomics research to analyze the most challenging low-abundance, high-complexity samples, to identify more compounds faster, quantify more accurately, and to elucidate structures more thoroughly.
Orbitrap Exploris Mass Spectrometers
Thermo Scientific Orbitrap Exploris mass spectrometry produces quantitative high-resolution, accurate-mass (HRAM) liquid chromatography mass spectrometry (LC-MS) with high quantitative accuracy and precision for your laboratory. You can meet your exact analytical requirements and translate your results more rapidly and simply than ever before with Orbitrap Exploris mass spectrometry.
Single Quadrupole IC-MS
Easily master MS for your IC applications.
Triple Quadrupole IC-MS
Need sensitive, fast, and robust quantitation of a variety of molecule types? For complex samples, the high sensitivity and selectivity of MS/MS using selected reaction monitoring (SRM) provides the most precise and accurate quantitation using IC-MS.
Orbitrap IC-MS
Thermo Scientific Orbitrap mass spectrometers generate high-resolution and accurate-mass data, thereby enhancing sensitivity in trace analyte detection, characterization, and quantitation. That's why they are used in research applications—in particular, life science applications such as metabolomics.
Get a superior combination of scan speed, high resolution, mass accuracy, spectral quality and sensitivity for untargeted or targeted screening and a broad range of qualitative and quantitative applications in environmental, food safety and metabolomics.
Use this system for IC-MS in life science metabolomics research to analyze the most challenging low-abundance, high-complexity samples, to identify more compounds faster, quantify more accurately, and to elucidate structures more thoroughly.
Orbitrap Exploris Mass Spectrometers
Thermo Scientific Orbitrap Exploris mass spectrometry produces quantitative high-resolution, accurate-mass (HRAM) liquid chromatography mass spectrometry (LC-MS) with high quantitative accuracy and precision for your laboratory. You can meet your exact analytical requirements and translate your results more rapidly and simply than ever before with Orbitrap Exploris mass spectrometry.
Which mass spectrometer do I choose?
Mass SpectrometerAnalyzerMode of OperationMSMS/MSSensitivityPeak ConfirmationStructural Elucidation
ISQ EC MSQuadrupoleSingle ion monitoring (SIM)
TSQ Altis MSTriple QuadrupoleSelected reaction monitoring (SRM)
TSQ Quantis MSTriple QuadrupoleSelected reaction monitoring (SRM)
Q Exactive MSOrbitrap mass analyzerHigh Resolution Accurate Mass (HRAM)
Orbitrap Fusion MSOrbitrap mass analyzerHigh Resolution Accurate Mass (HRAM)
Orbitrap Exploris MSOrbitrap mass analyzerHigh Resolution Accurate Mass (HRAM)
  = Not applicable  = Poor  = Good  = Better  = Best
IC-MS basics and benefits videos
Metabolomic Analysis using IC-MS
Prof. Izumi Yoshihiro talks about performing metabolomics analyses using IC-MS in his laboratory at Kyushu University in Japan.
Video Player is loading.
Current Time 0:00
Duration 1:48
Loaded: 0%
Stream Type LIVE
Remaining Time 1:48
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextMetabolomic Analysis using IC-MS
        Prof. Izumi Yoshihiro talks about performing metabolomics analyses using IC-MS in his laboratory at Kyushu University in Japan.
      2. Now Playing
        Up NextHow to Interface an Ion Chromatography System with a Mass Spectrometer
        Learn how to couple an ion chromatography to a mass spectrometric detector, including the importance of suppressor technology.
      3. Now Playing
        Up NextWhy Coupling Ion Chromatography with Mass Spectrometry Depends on Electrolytic Suppression
        Learn why electrolytic supression is critical to coupling ion chromatography with mass spectrometry.
      4. Now Playing
        Up NextThe Benefits of Using Ion Chromatography with Mass Spectrometry
        Frank Hoefler - Pittcon 2015 Seminar
      5. Now Playing
        Up NextPhytopharmaceutical analysis
        Hear how IC-MS is used at Ecocontrol Sud to analyze phytopharmaceuticals.
      6. Now Playing
        Up NextIC-MS for Water Safety
        Hear how Italy's Veritas uses IC-MS to detect highly polar pesticides in water.
    The Lab-Forward Trade-Up Program
    The world needs science to move faster. Now more than ever. Which is why when more scientists have access to the latest scientific instrumentation, we all benefit. That is the sole purpose of our Lab-Forward Trade-Up Program. For a limited time, trade up your chromatography and mass spectrometry instruments and save!
    Learn more
    The Ardia Platform is a scalable software solution for modern environments. It connects and organizes projects, people, and instruments, enabling scientists to collaborate more efficiently and gain new insights faster, helping them get closer to the truth.
    Learn more about Ardia Platform
    Resources