Sample Derivatization System
Automate derivatization and other small-scale reactions
Perform heating, stirring, and evaporation in a single system for a complete solution to conducting derivatization and other small-scale reactions with reliability and ease. The Thermo Scientific Reacti-Therm Sample Derivatization System can assist with gas chromatography (GC) sample preparation, which often includes small scale derivatization reactions such as silylation, alkylation, and acylation.
Complete solution for derivatization
The system includes the Thermo Scientific Reacti-Therm heating/stirring module for uniform dry heating of samples, which you can couple with the Reacti-Vap evaporator for simultaneous or separate delivery of pressurized gas.
  • Reliable and easy-to-use for constant temperatures
  • Uniform, stable dry heating: steady temperature incubation between ambient plus 10°C to 200°C
  • LED display: match digital display to in-block thermometer to calibrate temperature set-point
  • Modular design: switch aluminum blocks and vials; attach compatible evaporator manifold
  • In-block temperature control option by Remote Temperature Probe, an optional accessory to allow temperature regulation from block wells or actual sample vials
  • Magnetic stirrers for heating/stirring module for simultaneous stirring of samples in multiple vials
Customize the system to your needs
Reacti-Vials & Caps
Reacti-Vials & Caps
Convenient small sample handling
Reacti-Septum & Discs
Reacti-Septum & Discs
Inert, compatible with Reacti-Vials
Create your Reacti-Therm System
Download the brochure to review all the options available:
  • Heating, stirring and evaporation base units
  • Heated block sizes
  • Temperature control options
  • Stirrer bars and vials
 Download guide
Your go-to derivatization guide
Everything you need to know about derivatization
Download this 56 page derivatization guide about:
  • GC derivatization
  • HPLC ion pair derivatization
  • Amino acid analysis
  • Reagents and protocols
  • Sample handling
 Download guide
Need GC derivatization reagents?
CMD SchemaApp code