Intelligent
Built-in intelligence facilitates the full workflow from sample to results. Laboratories can be more reactive and profitable, with software that facilitates data driven decisions more efficiently.
Scientific 
Developed with scientific integrity, Qtegra ISDS Software delivers high quality, reliable data that labs can trust. Data transparency lets laboratory personnel focus on other tasks while ensuring the utmost confidence in scientific collaboration.
Data 
Data is the key to success for all laboratories. Qtegra ISDS Software can help labs excel at efficiently delivering consistent and relevant results with extensive tools to effectively acquire and manage data.
Solution 
Laboratories can enjoy the familiarity of common software across multiple analytical systems while differentiating their offering through customizable and application specific workflows.
Qtegra ISDS Software is compatible with a variety of powerful analytical instruments used to analyze trace elements and elemental isotopes. Use the table below to see what features are available for your application.
SupportedICP-OESQ-ICP-MSHR-IRMSMC-ICP-MSNoble gas MSGas IRMS
InstrumentsiCAP PRO ICP-OESiCAP MX Series ICP-MSUltra HR-IRMSNeoma MC-ICP-MSArgus VI, Helix SFT, Helix MCDELTA Q IRMS and 253 Plus 10 kV IRMS with peripherals
Get Ready automated set-up      
Intuitive and guided workflow      
21 CFR part 11 compliant environment  --- 
Autosampler  - - 
Autodilution  --- 
Desolvation nebulizers--- --
Chromatography integration-Optional using ChromControl Plug-in-Optional using ChromControl Plug-in- 
Single Cell analysis Optional using scQuant Plug-in----
Laser plug-in- -  -
Semi-quant analysisAxial only-----
Download the Intelligent Scientific Data Solution Software product specifications for more details, including:
  • Autosampler compatibility
  • Desolvation nebulizers
  • Laser plug-ins
Chromatography and ICP-MS Elemental Mass Spectrometry – Delivering New Insights
Determining sample safety or toxicity using ion chromatography (IC) coupled with inductively coupled plasma mass spectrometry (IC-ICP-MS).
Video Player is loading.
Current Time 0:00
Duration 3:15
Loaded: 0.00%
Stream Type LIVE
Remaining Time 3:15
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up NextChromatography and ICP-MS Elemental Mass Spectrometry – Delivering New Insights
        Determining sample safety or toxicity using ion chromatography (IC) coupled with inductively coupled plasma mass spectrometry (IC-ICP-MS).
      2. Now Playing
        Up NextClumped carbonate isotope analysis driven by Qtegra ISDS Software
        Explore innovative data evaluation setup for ultimate accuracy.
      3. Now Playing
        Up NextScientific Research with Qtegra ISDS Software
        Gain flexibility to tackle the most demanding applications for your research.
      4. Now Playing
        Up NextAnalytical Science with Qtegra ISDS Software
        Power your lab productivity and bring simplicity in your routine analysis.
      5. Now Playing
        Up NextEA-IRMS driven by Qtegra ISDS Software
        Discover simplified workflows for boosting your EA-IRMS analysis.
      6. Now Playing
        Up NextGC-IRMS driven by Qtegra ISDS Software
        Perfect your GC-IRMS research with our flexible and fully transparent software platform.
      7. Now Playing
        Up NextGas IRMS with Qtegra ISDS Software
        The journey has begun.
      8. Now Playing
        Up NextGlobal pharmaceutical CRO chooses iCAP RQ & Qtegra ISDS software for identification and quantification of elemental Extractable & Leachable impurities
        Elemental analysis and trace metals testing are important for the commercial release of pharmaceutical products, stability studies, extractable and leachable studies, and clinical trial support.
    Webinars
    Video Player is loading.
    Current Time 0:00
    Duration 45:02
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 45:02
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Qtegra ISDS Software for gas IRMS webinar: revolutionize your gas IRMS analyses
      Video Player is loading.
      Current Time 0:00
      Duration 1:04:28
      Loaded: 0.00%
      Stream Type LIVE
      Remaining Time 1:04:28
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Learn how Intelligent Software can streamline your elemental analysis workflow
        Video Player is loading.
        Current Time 0:00
        Duration 1:00:09
        Loaded: 0.00%
        Stream Type LIVE
        Remaining Time 1:00:09
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Exploring trace element intracellular processes at the single cell level using ICP-MS
          * Required field
          *
          *
          *
          *
          *
          *
          *
          *
          *
           Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
          This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
          CMD Wide-format style fixes