Antibody purification involves isolation of antibody from serum (polyclonal antibody), ascites fluid, or from the culture supernatant of a hybridoma cell line (monoclonal antibody). Protein L is a recombinant protein expressed in Escherichia coli (35.8 kDa; 4 IgG binding sites) and is recommended for human or mouse monoclonal antibodies known to have appropriate kappa light chains.
We offer two base supports and a variety of formats to purify antibodies using the Protein L ligand. Our portfolio is designed to meet small-scale (screening) to pilot-scale needs.
  • Multiple formats—magnetic beads, loose resin, spin columns and kits, and FPLC cartridges enable antibody purification from microgram to kilogram scales
  • High performance—resins are designed to maximize protein yield and reduce background
  • Economical—pricing is similar to or better than our leading competitors
Choose the right Protein L product for antibody purification
Product links in the last row of each column
 Pierce Protein L
Magnetic Beads
Pierce Protein L
Agarose
Pierce Protein L Plus
Agarose
Bead or resin size1 μm45–165 μm45–165 μm
Binding capacity≥110 μg human IgG/mg beads5–10 mg human IgG/mL resin10–20 mg human IgG/mL resin
Maximum linear flow rateN/A700 cm/hr700 cm/hr
SupportMagnetite coated polymerCrosslinked 6% beaded agaroseCrosslinked 6% beaded agarose
Recommended application scalescreeningbatch, pilotbatch, pilot
Formats availableloose beadsloose beads, pre-packed spin columns and kits, chromatography cartridgesloose beads
Order product(s)8884920510
(loose resin)
89928
(1 mL cartridge)
89929
(5 mL cartridge)
20520
(loose resin)
Protein L spin columns and spin kits
Catalog # Name Size Price (USD) Qty
89955 NAb™ Protein L Spin Columns, 0.2 mL Each
502.00
89959 NAb™ Protein L Spin Columns, 1 mL Each
709.65
Online Exclusive
946.00
Save 236.35 (25%)
89963 NAb™ Protein L Spin Column, 5mL Each
934.00
89951 NAb™ Protein L Spin Kit, 0.2 mL Each
552.00
89981 NAb™ Protein L Spin Kit, 1 mL Each
459.00
21189 Pierce™ Recombinant Protein L, 1 mg Each
122.65
Online Exclusive
135.00
Save 12.35 (9%)
For Research Use Only. Not for use in diagnostic procedures.