Antibody purification involves isolation of antibody from serum (polyclonal antibody), ascites fluid, or from the culture supernatant of a hybridoma cell line (monoclonal antibody). Protein G is a recombinant protein expressed in Escherichia coli (21.6 kDa; 2 IgG binding sites) and is recommended for purifying IgG from mouse, human, cow, goat, or sheep serum. The POROS MabCapture G Select Resin is a product that can be scaled up for bioprocess applications.
We offer a wide selection of base supports and formats to purify antibodies using the Protein G ligand. Our portfolio is designed to meet small-scale (screening) up to large-scale (bioprocess) needs.
  • Multiple formats—loose resin, spin columns and kits, FPLC cartridges, and 96-well filter plates enable antibody purification from microgram to kilogram scales
  • High performance—resins are designed to maximize protein yield and reduce background
  • Economical—pricing is similar to or better than our leading competitors
Choose the right Protein G product for antibody purification
Product links in the last row of each column
 Pierce Protein G AgarosePierce Protein G Plus AgarosePOROS MabCapture G Select
Bead or resin size45–165 μm45–165 μm50 μm
Binding capacity11–15 mg human IgG/mL resin≥20 mg human IgG/mL resin≥13 mg/mL human IgG/mL resin
Maximum linear flow rate700 cm/hr700 cm/hr3000 cm/hr
SupportCrosslinked 6% beaded agaroseCrosslinked 6% beaded agarosePolystyrene-divinylbenzene
Recommended application scalebatch, pilotbatch, pilotbatch, pilot, process
Formats availableloose beads, pre-packed spin columns and kits, chromatography cartridges, spin platesloose beadsloose beads
Order product(s)20398
(loose resin)
89926
(1 mL cartridge)
89927
(5 mL cartridge)
45204
(spin plates)
22851
(loose resin)
82082
Protein G spin columns, spin kits, and ready-to-use purification buffers
Catalog # Name Size Price (USD) Qty
89953 NAb™ Protein G Spin Columns, 0.2 mL Each
415.00
89957 NAb™ Protein G Spin Columns, 1 mL Each
854.00
89961 NAb™ Protein G Spin Columns, 5 mL Each
782.00
89949 NAb™ Protein G Spin Kit, 0.2 mL Each
462.00
89979 NAb™ Protein G Spin Kit, 1 mL Each
374.00
21193 Pierce™ Recombinant Protein G, 5 mg Each
235.65
Online Exclusive
274.00
Save 38.35 (14%)
21019 Pierce™ Protein G IgG Binding Buffer, 1 L Each
108.65
Online Exclusive
124.00
Save 15.35 (12%)
21004 Pierce™ IgG Elution Buffer, 1 L Each
112.65
Online Exclusive
129.00
Save 16.35 (13%)
21020 Pierce™ Gentle Ag/Ab Binding Buffer, pH 8.0, 1 L Each
184.00
21027 Pierce™ Gentle Ag/Ab Elution Buffer, pH 6.6, 500 mL Each
154.65
Online Exclusive
175.00
Save 20.35 (12%)
21030 Pierce™ Gentle Ag/Ab Binding and Elution Buffer Kit Each
178.00
For Research Use Only. Not for use in diagnostic procedures.