Search Thermo Fisher Scientific
CaptureSelect protein purification products exhibit superior affinity and specificity for their target proteins, for efficient single-step purification of non-antibody biomolecules. In addition, sensitive proteins can be efficiently eluted using mild conditions. These protein purification resins were designed for reliable, efficient purification of target proteins, with essential stability and elution properties.
CaptureSelect protein purification products offer:
Product | Characteristics | BioProcess |
---|---|---|
POROS CaptureSelect AAV8 | For purification of Adeno-Associated Virus Type 8 (AAV8) | AAV8 |
POROS CaptureSelect AAV9 | Specific for Adeno-Associated Virus Type 9 (AAV9) | AAV9 |
POROS CaptureSelect AAVX | For the purification of native and recombinant adeno-associated virus particles (AAV) of various serotypes | AAVX |
POROS CaptureSelect AdV5 | For purification and/or scavenging of recombinant hexon capsomeres of Adenovirus serotype 5 (AdV5) particles as well as free hexon proteins thereof | |
CaptureSelect Follicle Stimulating Hormone | Purifies intact Follicle Stimulating Hormone (FSH) | Follicle Stimulating Hormone |
CaptureSelect hCG | Binds to the alpha chain of HCG (human chorionic gonadotropin) | hCG |
CaptureSelect Human Albumin | Specific for human serum albumin, suitable for excipient grade HSA and HSA-fusion proteins | HSA and HSA-fusion proteins |
CaptureSelect Multi-Species Albumin Depletion Product | Depletes albumin from plasma or serum derived from a number of different species (e.g., mouse, rat, rabbit, goat, bovine, guinea pig, human, and horse) | |
CaptureSelect tPA | Suitable for human tissue Plasminogen Activator (tPA) | tPA |
CaptureSelect Human Growth Hormone | Works for both CHO as well as E. coli derived hGH | hGH |
CaptureSelect FactorII(a) | Purifies plasma and recombinant Factor II and Factor IIa | |
CaptureSelect Factor H | Suitable for human complement Factor H from plasma and recombinant sources | |
CaptureSelect Antithrombin III | Both alpha as well as beta Antithrombin III (ATIII) are bound | |
CaptureSelect AAT-XL | Purifies plasma and recombinant Alpha-1 Antitrypsin (AAT) | AAT-XL |
CaptureSelect Factor X | Purifies Factor X from plasma and recombinant sources | Factor X |
POROS CaptureSelect Fib XL | Improved version of CaptureSelect Fibrinogen Affinity Matrix with an increased binding capacity. Suitable for plasma derived and recombinant Fibrinogen. | |
CaptureSelect Transferrin | Suitable for plasma derived and recombinant Transferrin | |
CaptureSelect TSH | For the purification of thyroid-stimulating hormone (TSH) from recombinant sources | TSH |
POROS CaptureSelect C1-InhXL | Purifies plasma and recombinant human C1 esterase inhibitor | C1-InhXL |
CaptureSelect GM-CSF | Suitable for recombinant human Granulate-macrophage colony-stimulating factor (GM-CSF) |
Check out the new Chromatography Learning Lab from Thermo Fisher Scientific, providing a robust range of webinars, articles, eBooks, infographics, and other digital resources to help you optimize the use of our purification products in your process.
Explore digital resource
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
For Research Use Only. Not for use in diagnostic procedures.