Fill finish solution
Maximize drug product recovery
The Thermo Scientific Fill Finish solution offers a complete, configurable solution for BioPharma and CDMOs looking to maximize drug product recovery. With industry-leading integrity assurance, excellent lead times, and a range of easy-to-use, single-use consumables for compounding, sterile filtration (PUPSIT), and aseptic filling applications, you can enable high product quality while optimizing your workflow. Customizable and engineered-to-order, the Fill Finish Solution provides a complete solution to help finish application needs.
Contact us  Download data sheet
Fill Finish Solution Aseptic Filling Assembly: 3D product tour
Check out this interactive tool to learn more about how our fill finishing solution can help maximize drug product recovery and offer exceptional lead times with a customizable, configurable system.
Single-use assemblies featuring:
Designed with maximizing drug product recovery in mind, the Thermo Scientific Fill Finish Solution offers key features:
  • Maximum product recovery and optimal drainage with Labtainer Pro BioProcess Container design
  • Minimize the risk of leaks and simplify set up with BioTitan Retention Device
  • 100% assembly helium integrity test with 10 μm detection limit
  • Exceptional lead times due to standard and configurable product design 
  • Proven, high-quality film featuring leading single-use Aegis 5-14 film 
Close up of clear bag with red filling and red tubing
The Fill Finish product family provides configurable, single-use assemblies from a standard component catalog and standard assemblies for drug product final fill application. Products include compounding assemblies, sterile filtration, and aseptic filling assemblies.
1743584485998.jpg
Compounding assembly
  • Buffer to compounding BPC
  • Compounding to holding BPC
  • Bioburden reduction filter assembly
Filtration assembly
  • PUPSIT
  • Double sterilization filter assembly
Filling assembly
  • Beta bag
  • Drug surge bag
  • Tubing assembly
  • Filling needle
Watch this video to learn more
Video Player is loading.
Current Time 0:00
Duration 2:16
Loaded: 0%
Stream Type LIVE
Remaining Time 2:16
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Related products
    Bioprocessing resources
    Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
    For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.