Find the right Attune Flow Cytometer model for your lab
All of the Attune Flow Cytometers are compact benchtop cell-analyzers that utilize acoustic focusing technology and share common features including exceptionally fast sample flow rates, high clog resistance, and outstanding reliability. Use the comparison guide below to compare the Attune NxT Flow Cytometer with the Attune CytPix Flow Cytometer and the Attune Xenith Flow Cytometer so that you can select the right model for your research needs.
Which Attune Flow Cytometer is right for you?
Attune Xenith Flow Cytometer
Attune CytPix Flow Cytometer
Attune NxT Flow Cytometer
Product summary Attune Flow Cytometer with advanced spectral capabilities including UV and NIR lasers, supporting both traditional compensation and spectral unmixing analysis. Attune Flow Cytometer with a high-resolution brightfield camera and automated image analysis software for morphometric analysis in combination with standard fluorescence and scatter parameters. Advanced, high throughput, acoustic focusing, benchtop flow cytometer with up to 14 color/16 parameter flow cytometry analysis.
Applications Spectral high-parameter analyses
  • New users
  • Cell and gene therapy
  • Quality control
  • New users
  • High throughput, standard parameter analysis
  • Rare event detection
Feature
Optics (fluorescence detection)
Number of lasers
6 (fixed) 2–4 (see table below) 1–4 (see table below)
Number of detection channels/emission filters 57 channels with wavelength-tuned photomultiplier tubes (PMTs) (51 fluorescence channels); user-changeable, keyed filters 2 scatter channels, up to 14 color channels with wavelength-tuned photomultiplier tubes (PMTs); user-changeable, keyed filters 
Optical alignment
Fixed alignment with pre-aligned welded fiber; no user maintenance required
Optics (imaging) Laser excitation N/A 405 nm laser with <50 nanosecond pulse width N/A
Pulse width N/A <50 ns N/A
Fluidics
Acoustic focusing Yes Yes Yes
Custom sample flow rates Yes Yes Yes
Performance (fluorescence detection)
Fluorescence sensitivity ≤30 MESF for FITC, ≤10 MESF for PE, ≤10 MESF for APC ≤80 MESF for FITC, ≤30 MESF for PE, ≤70 MESF for APC
Fluorescence resolution rCV <3% for the singlet peak of propidium iodide–stained chicken erythrocyte nuclei (CEN) CV <3% for the singlet peak of propidium iodide-stained chicken erythrocyte nuclei (CEN)
Data acquisition rate
Up to 35,000 events/sec with all parameters
Maximum electronic speed >100,000 events/sec with all parameters 65,000 events/sec with all parameters
Performance (imaging)
Image capture rate N/A Up to 6,000 images/sec depending on image size and event rate N/A
Objective N/A Magnification 20x, N/A
Numerical aperture 0.45
Pixel resolution N/A 0.3 microns/pixel N/A
Detection limit N/A Visually detect 800 nm particles N/A
Image analysis software Models N/A Thermo Fisher–supplied models: leukocytes, beads; User-trained models: based on sample or particle type N/A
Parameters/Features N/A 36 measured morphology parameters in 10 categories: shape features, intensity and texture features, object features, pixel features, particle interaction, particle count, similarity calculation, moment weighted, co-occurrence and system features  N/A
Sample size range of models N/A 5–20 µm N/A
Image processing speed N/A 1,000 images/second
(varies by image size and complexity)
N/A
Spectral analysis software Spectral unmixing Spectral unmixing with autofluorescence subtraction and live unmixing, and use of tubes and wells N/A N/A
Quality and regulatory
Instrument tracking Automated daily baseline and performance test with Levey-Jennings plots
Regulatory status For Research Use Only
Physical
Dimensions (H x W x D) including fluid bottles ~50 x 86 x 61 cm (~19.5 x 34 x 24 in) ~49 x 58 x 43 cm (~19 x 23 x 17 in) ~40 x 58 x 43 cm (~16 x 23 x 17 in)
Biosafety hood compatibility Yes Yes Yes
Computer
Software requirements LSOne Software Invitrogen Attune Cytometric Software
Memory 64 GB 64 GB (4 x 16 GB) DDR4 2666 MHz UDIMM Non-ECC 32 GB
Hard drives 2 x 8 TB SSD, 560 MB/sec; controller RAID1, integrated 2 x 8 TB SSD, 2.5-inch Samsung 870 QVO, 560 MB/s 2 x 2 TB SATA 3.0 GB/s, 8MB data burst cache; Controller RAID1, integrated
Graphics processor NVIDIA RTX A2000 Nvidia Quadro P2200 N/A
Laser–detector options for Attune Nxt and CytPix cytometers
Lasers Laser configuration Number of detection channels for included lasers Total detection channels* Attune CytPix Attune NxT
Violet, 405nm Blue, 488nm Yellow, 561nm Green, 532nm Red, 637nm
1 Blue
4 6 N/A A24864
2 Blue/green
3 4 9 N/A A28995
Blue/yellow
3 4 9 A51842 A24861
Blue/red
4 3 9 A51840 A24863
Blue/violet
4 4 10 A51841 A24862
Blue/violet 6
6 3 11 A51843 A29002
3 Blue/green/red
3 4 3 12 N/A A28997
Blue/red/yellow
3 4 3 12 A51845 A28993
Blue/green/violet
4 3 4 13 N/A A28999
Blue/violet/yellow
4 3 4 13 A51846 A24859
Blue/red/violet
4 4 3 13 A51844 A24860
Blue/red/violet 6
6 3 3 14 A51847 A29003
4 Blue/red/violet/green
4 3 4 3 16 N/A A29001
Blue/red/yellow/violet
4 3 4 3 16 A51848 A24858
Blue/red/yellow/violet 6
6 2 3 3 16 A51849 A29004
Laser power specifications for Attune Flow Cytometers
For Research Use Only. Not for use in diagnostic procedures.
1x1 image pixel for data collection1x1 image pixel for data collection