Efficient, flexible, transformative flow cytometer with up to 14 colors
The Attune NxT Flow Cytometer is an advanced cell analyzer in which acoustic focusing fluidics is the key to both high sensitivity and high throughput.
Features
The Attune NxT Flow Cytometer shares common features with the Attune CytPix model.
Acoustic focusing for sensitivity
Fast, accurate acquisition
Rare event detection
Designed for flexibility
Novel optical design
Clog-resistant technology
Powerful, intuitive software
Biosafety hood compatible
For our imaging-enhanced flow cytometer, check out the Attune CytPix model.
Flow cytometry applications
Almost any standard flow cytometry application can benefit from the high sensitivity and throughput of the Attune NxT Flow Cytometer. See our Sample Data page for examples spanning infectious disease research, fluorescent proteins, CRISPR gene editing, immuno-oncology research, microbiology, plant ploidy, platelets, stem cells, and T cells.
One classic flow cytometry application is immunophenotyping. Flow cytometry is the method of choice for identifying cells within complex heterogeneous populations, as it allows for multiparameter analysis of thousands to millions of cells in a short time. Strong signal separation in the Attune NxT Flow Cytometer shows excellent separation of cell populations into subsets for immunophenotyping. A wide range of reagent choices—as well as the system’s automated compensation module, four spatially separated lasers, and 14 color choices—help to simplify multicolor panel design.
Gating strategy for 13-color immunophenotyping analysis of stained human whole blood using a stain/lyse protocol. Human whole blood cells were stained as described in the application note and acquired and analyzed on the Attune NxT Flow Cytometer. (A) Dead cells were excluded from the analysis by gating on live (propidium iodide) cells in a dot plot. (B) CD45+ gating was used to select the leukocyte population from the lysed whole blood. (C) Lymphocytes and monocytes were identified based on forward and side scatter profiles. (D) Monocytes are found above lymphocytes on the scatter plot and express both CD14 and CD33. (F) Within the lymphocyte gate, immune cells can be subdivided based on their expression of CD3 (T cells), CD19 (B cells), or neither (NK cells). (E) B cells can be further characterized by HLA-DR and CD45RA expression. (G) T cells can be further subdivided into CD4+ (T helper cells) and CD8+ (cytotoxic T cells) subpopulations, while (J) regulatory T cells express CD4 and CD25. (H, K) CD62L identifies naive (TN) CD4 and CD8 T cells, while HLA-DR is expressed by activated T cells (TA). (I) Finally, NK cells lack B cell and T cell markers (CD19CD3) and express CD56.
Videos and demos
Laser and detector configurations
Lasers Laser configuration Lasers included, No. of detection channels Total detection channels* Category Number
Violet, 405nm Blue, 488nm Yellow, 561nm Green, 532nm Red, 637nm
1 Blue
4 6 A24864
2 Blue/green
3 4 9 A28995
Blue/yellow
3 4 9 A24861
Blue/red
4 3 9 A24863
Blue/violet
4 4 10 A24862
Blue/violet 6
6 3 11 A29002
3 Blue/green/red
3 4 3 12 A28997
Blue/red/yellow
3 4 3 12 A28993
Blue/green/violet
4 3 4 13 A28999
Blue/violet/yellow
4 3 4 13 A24859
Blue/red/violet
4 4 3 13 A24860
Blue/red/violet 6
6 3 3 14 A29003
4 Blue/red/violet/green
4 3 4 3 16 A29001
Blue/red/yellow/violet
4 3 4 3 16 A24858
Blue/red/yellow/violet 6
6 2 3 3 16 A29004
Specifications
Optics: Fluorescence detection
Laser power
Laser
Wavelength (nm)
Beam-shaping optics (BSO)* (mW)
Diode power** (mW)
Violet
405
50
100
Blue
488
50
100
Green
532
100
140
Yellow
561
50
100
Red
637
100
140
* Amount of measured usable laser power after light has gone through the beam optics and shaping filters.
** Vendor-specified theoretical maximum.
Laser excitation
Optimized excitation for minimized stray laser-line noise and losses to reflection
Laser profile
10 x 50 μm flat-top laser providing robust alignment
Emission filters
Up to 14 color channels with wavelength-tuned photomultiplier tubes (PMTs); user-changeable, keyed filters
Laser separation
150 μm
Optical alignment
Fixed alignment with prealigned welded fiber; no user maintenance required
Onboard thermoelectric cooler
No warm-up delay; fiber isn’t affected by “on/off”
Simmer mode
Instant “on/off” reduces usage and/or aging by 10x; only turns on when acquiring samples; reports hours of usage
Flat-top laser specified at the flow cell
Coefficient of variation (CV) <3% over the width of the flat-top laser
Upgradable
Convenient field changes
Fluidics
Flow cell
Quartz cuvette gel coupled to 1.2 numerical aperture (NA) collection lens, 200 x 200 μm
Sample analysis volume
20 μL–4 mL
Custom sample flow rates
12.5–1,000 μL/min
Sample delivery
Positive-displacement syringe pump for volumetric analysis
Sample tubes
Accommodates tubes from 17 x 100 mm to 8.5 x 45 mm
Fluid-level sensing
Active
Standard fluid reservoirs
1.8 L focusing fluid tank, 1.8 L waste tank, 175 mL shutdown solution tank, and 175 mL wash solution tank
Fluid storage
All fluids stored within instrument
Extended fluidics option
Configuration for 10 L fluid
Nominal fluid consumption
1.8 L/day
Automated maintenance cycles
≤15 min start-up and shutdown—deep clean, sanitize, and debubble modes
Performance: Fluorescence detection
Fluorescence sensitivity
≤80 molecules of equivalent soluble fluorochrome (MESF) for FITC, ≤30 MESF for PE, ≤70 MESF for APC
Fluorescence resolution
CV <3% for the singlet peak of propidium iodide–stained chicken erythrocyte nuclei (CEN)
Data acquisition rate
Up to 35,000 events/sec, 34 parameters, based on a 10% coincidence rate per Poisson statistics
Maximum electronic speed
65,000 events/sec with all parameters
Carryover
Single-tube format: <1%
Forward and side scatter sensitivity
Able to discriminate platelets from noise
Forward and side scatter resolution
Optimized to resolve lymphocytes, monocytes, and granulocytes in lysed whole blood
Forward scatter
Photodiode detector with 488/10 nm bandpass filter
Side scatter
PMT with default 488/10 nm bandpass filter; optional 405/10 nm bandpass filter
Fluorescence detectors
14 individual detectors
Electronic pulse
Measured area, height and width pulse for all detectors
Violet side scatter resolution
Can be configured for violet side scatter to better resolve particles from noise
Minimum particle size
0.2 μm on side scatter using submicron bead calibration kit from Bangs Laboratories—0.1 μm on side scatter under following conditions: Using an Attune NxT Flow Cytometer with standard 0.5 mm blocking configuration, an Invitrogen Attune NxT 488/10 Filter (Cat. No. 100083194), and Attune Focusing Fluid (Cat. No. 4488621, 4449791, or A24904) that has been passed through a 0.025 µm filter
Software
Compensation
Full matrix—automated and manual modes, on-plot compensation tools for fine adjustment; use of tubes and wells
Flow rate
Precise flow rate control via software; no hardware adjustments
Live streaming
Live update of statistics during acquisition of events up to 35,000 events/sec
Overlays
Comparative analysis between samples; 3D view
Sample recovery
System able to return unused samples
Concentration
Direct concentration measurement without use of counting beads
Software layout
Fully customizable for each user account
Bubble detection technology
Stops automated run to preserve sample integrity
Maximum single-event file
20 million with option to append
Heat map
Set up for definition of plate layout; screening view for analysis for tubes and plates
Threshold
Up to 4 individual thresholds with user option to apply Boolean logic
Gating
Hierarchical gating with the ability to derive gates
Voltage
User adjustable
Window extensions
User adjustable
Area scaling factor (ASF)
User adjustable
Acquisition settings
Documented in FCS files and maintained upon import
Templates
Create from existing experiments—instrument settings, workspaces, run protocols, heat map settings, and compensation settings optimized and defined previously
Tube-to-plate conversion
One-click transition from tubes to plates and vice versa; no disassembly, no additional QC, no reboot required for conversion between plates and tubes
Graphics resolution
Publication-quality images; support for TIF, PNG, BMP, JPG, GIF, and EMF; quickly copy and paste plots to any external application (e.g., Microsoft™ PowerPoint™ software)
User account administration
Administrative creation of individual user accounts with designated roles, advanced setting permissions, management of individual accounts, user time tracking, and sample count
Quality and regulatory
Instrument tracking
Automated daily baseline and performance test with Levey-Jennings plots
Warranty
1 year
Production verification testing
Each instrument is tested and verified for assembly integrity and performance to specifications
Quality management system
Manufacturing standards comply with the requirements of ISO 13485:2003
Robust installation specifications
Units installed by engineer; preplanning checklist, delivery, and installation; and performance validation compliance with standardized procedure
Regulatory status
For Research Use Only
Computer
Software requirements
Invitrogen Attune Cytometric Software
Monitor
23 in. flat panel (1,920 x 1,080 resolution); dual-monitor capability
Computer
Minitower desktop
Operating system
Microsoft™ Windows™ 10 64-bit
FCS format
FCS 3.1, 3.0
Processor
Intel™ Core™ i7 processor
RAM
32 GB
Hard drives
2 x 2 TB SATA 3.0 Gb/s, 8 MB data burst cache; controller RAID1, integrated
Installation requirements
Electrical requirements
100–240 VAC, 50/60 Hz, <150 W
Thermo Fisher Scientific certifies that the Attune Flow Cytometers conform to relevant directives to bear the CE mark. The instrument also conforms to the UL and CAN/CSA general requirements (61010.1). The Attune Flow Cytometers are Class I laser products per Center for Devices and Radiological Health (CDRH) regulations and EN/IEC 60825.
Heat dissipation
<150 W
Temperature operating ranges
15–30°C (59–86°F)
Operating humidity
10–90%, noncondensing
Audible noise
<65 dBA at 1.0 m
Instrument size (H x W x D)
~40 x 58 x 43 cm (16 x 23 x 17 in.), including fluid bottles
Weight
~29 kg (64 lb)