KingFisher Ready DNA Ultra 2.0 Prefilled Plates
Introducing, pre-filled plates for easier, faster, and more consistent DNA extractions. Simply peel off cover, add lysed sample and load the plate on KingFisher.
  • MagMAX DNA reagents prefilled into KingFisher plates
  • Select Duo-Ready or Flex-Ready kits for your KingFisher instruments    
Buy Flex-readyBuy Duo-readyWatch video
 KingFisher Flex-Ready DNA Ultra 2.0 Prefilled plates
KingFisher Flex-Ready DNA Ultra 2.0 Prefilled plates
Process 96 samples in 51 minutes – without buffer dispensing time
KingFisher Duo-Ready DNA Ultra 2.0 Prefilled plates
KingFisher Duo-Ready DNA Ultra 2.0 Prefilled plates
Process 12 samples in 45 minutes – without buffer dispensing time
Sample input volume50–400 mL50–400 mL
Samples processed in kit96 samples96 samples
Sample input typesWhole blood, saliva, buccal swabs, buffy coat, blood cards, tissue culture cellsWhole blood, saliva, buccal swabs, buffy coat, blood cards, tissue culture cells
KingFisher run time51 min45 min
All accessory plastics
  • All necessary 6 sealed and prefilled Flex 96 deep well plates
  • 8 sealed, prefilled Flex 96 deep well plates
  • Flex deck position printed on each plate
  • Flex 96-tip comb in protective holder
  • Color-coded bottles
  • 8 sealed, prefilled Duo 96 deep well plates
  • 8 Duo 12-tip combs
  • Elution strips + caps
  • Elution strip frame
  • Color-coded bottles
Order now
Order now
workflow
Easy workflow for fewer errors and less worry
  • Peel off foil seals, add enhancer, sample and Proteinase K solutions, insert plate(s) into instrument
  • Eliminates the need to dispense lysis or wash solutions or magnetic beads
  • Tip combs, sample plates, and elution strips/caps conveniently included in the box
  • Reduce errors associated with complicated plate maps
Same sample flexibility, yield and quality as MagMAX Ultra 2.0
  • Flexible: One kit will process multiple sample input types
  • Scalable: Process from 50 – 400 ul of sample input; no need to adjust reagents
  • Automatable: Magnetic bead based chemistry optimized for KingFisher Instruments
  • Compatibility: Get high yield and high quality nucleic acid suitable for multiple downstream applications
MagMAX performance in KingFisher plates
Blood
Linear recovery of DNA across a wide range of whole blood sample volume inputs
Figure 1. Linear recovery of DNA across a wide range of whole blood sample volume inputs.
Linear recovery of DNA from buffy coat collected with K2 EDTA blood collection tube and Streck cfDNA BCT
Figure 2. Linear recovery of DNA from buffy coat collected with K2 EDTA blood collection tube and Streck cfDNA BCT.
Buffy Coat
Greater yields and purity obtained with MagMAX Ultra 2.0
Figure 3. Greater yields and purity obtained with MagMAX Ultra 2.0.
Purity of the isolated DNA is very high with A260/A280 ratios A260/A230 ranges above 1.8 indicating the high purity of the isolated DNA
Figure 4. Purity of the isolated DNA is very high with A260/A280 ratios A260/A230 ranges above 1.8 indicating the high purity of the isolated DNA.
How to use KingFisher Duo-Ready DNA Ultra 2.0 prefilled plates for KingFisher Duo Prime
Process 96 samples with KingFisher Duo-Ready DNA Ultra 2.0 prefilled plates for KingFisher Duo Prime. This will show you how to process blood samples. A similar process can be used for other sample types.
Video Player is loading.
Current Time 0:00
Duration 4:05
Loaded: 0%
Stream Type LIVE
Remaining Time 4:05
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    How to use KingFisher Flex-Ready DNA Ultra 2.0 prefilled plates for KingFisher Flex
    Process 96 samples with KingFisher Flex-Ready DNA Ultra 2.0 prefilled plates for KingFisher Flex. This will show you how to process blood samples. A similar process can be used for other sample types.
    Video Player is loading.
    Current Time 0:00
    Duration 4:32
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 4:32
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
       KingFisher Flex-Ready DNA Ultra 2.0 Prefilled plates
      KingFisher Flex-Ready DNA Ultra 2.0 Prefilled plates
      Process 96 samples in 51 minutes – without buffer dispensing time
      KingFisher Duo-Ready DNA Ultra 2.0 Prefilled plates
      KingFisher Duo-Ready DNA Ultra 2.0 Prefilled plates
      Process 12 samples in 45 minutes – without buffer dispensing time
      Sample input volume50–400 mL50–400 mL
      Samples processed in kit96 samples96 samples
      Sample input typesWhole blood, saliva, buccal swabs, buffy coat, blood cards, tissue culture cellsWhole blood, saliva, buccal swabs, buffy coat, blood cards, tissue culture cells
      KingFisher run time51 min45 min
      All accessory plastics
      • All necessary 6 sealed and prefilled Flex 96 deep well plates
      • 8 sealed, prefilled Flex 96 deep well plates
      • Flex deck position printed on each plate
      • Flex 96-tip comb in protective holder
      • Color-coded bottles
      • 8 sealed, prefilled Duo 96 deep well plates
      • 8 Duo 12-tip combs
      • Elution strips + caps
      • Elution strip frame
      • Color-coded bottles
      Order now
      Order now
      workflow
      Easy workflow for fewer errors and less worry
      • Peel off foil seals, add enhancer, sample and Proteinase K solutions, insert plate(s) into instrument
      • Eliminates the need to dispense lysis or wash solutions or magnetic beads
      • Tip combs, sample plates, and elution strips/caps conveniently included in the box
      • Reduce errors associated with complicated plate maps
      Same sample flexibility, yield and quality as MagMAX Ultra 2.0
      • Flexible: One kit will process multiple sample input types
      • Scalable: Process from 50 – 400 ul of sample input; no need to adjust reagents
      • Automatable: Magnetic bead based chemistry optimized for KingFisher Instruments
      • Compatibility: Get high yield and high quality nucleic acid suitable for multiple downstream applications
      MagMAX performance in KingFisher plates
      Blood
      Linear recovery of DNA across a wide range of whole blood sample volume inputs
      Figure 1. Linear recovery of DNA across a wide range of whole blood sample volume inputs.
      Linear recovery of DNA from buffy coat collected with K2 EDTA blood collection tube and Streck cfDNA BCT
      Figure 2. Linear recovery of DNA from buffy coat collected with K2 EDTA blood collection tube and Streck cfDNA BCT.
      Buffy Coat
      Greater yields and purity obtained with MagMAX Ultra 2.0
      Figure 3. Greater yields and purity obtained with MagMAX Ultra 2.0.
      Purity of the isolated DNA is very high with A260/A280 ratios A260/A230 ranges above 1.8 indicating the high purity of the isolated DNA
      Figure 4. Purity of the isolated DNA is very high with A260/A280 ratios A260/A230 ranges above 1.8 indicating the high purity of the isolated DNA.
      How to use KingFisher Duo-Ready DNA Ultra 2.0 prefilled plates for KingFisher Duo Prime
      Process 96 samples with KingFisher Duo-Ready DNA Ultra 2.0 prefilled plates for KingFisher Duo Prime. This will show you how to process blood samples. A similar process can be used for other sample types.
      Video Player is loading.
      Current Time 0:00
      Duration 4:05
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 4:05
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        How to use KingFisher Flex-Ready DNA Ultra 2.0 prefilled plates for KingFisher Flex
        Process 96 samples with KingFisher Flex-Ready DNA Ultra 2.0 prefilled plates for KingFisher Flex. This will show you how to process blood samples. A similar process can be used for other sample types.
        Video Player is loading.
        Current Time 0:00
        Duration 4:32
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 4:32
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • en (Main), selected
          Video: KingFisher Ready DNA Ultra 2.0 prefilled plates
          KingFisher Ready DNA Ultra 2.0 prefilled plates are ready-to-use reagents using MagMAX technology, which means fewer pipetting steps and faster, more consistent extractions. Simply peel off cover, add sample, and load plate onto a KingFisher.
          Video Player is loading.
          Current Time 0:00
          Duration 1:38
          Loaded: 0%
          Stream Type LIVE
          Remaining Time 1:38