Cell-free DNA (cfDNA) refers to fragments of DNA that are found circulating freely in the bloodstream or other bodily fluids. Types of cfDNA include circulating tumor DNA (ctDNA), cell-free fetal DNA (cffDNA), cell-free mitochondrial DNA (ccf mtDNA), and donor-derived cell-free DNA (dd-cfDNA).
Efficient nucleic acid isolation that specifically recovers the circulating genetic material containing information related to the disease state is the first step in attaining valuable information. The importance of cfDNA is becoming established, but it may not tell the whole story. Cell-free RNA (cfRNA) sequences may serve as biomarkers as well in the form of microRNAs and gene fusions. Inclusion of cfRNA with cfDNA research provides an opportunity for researchers to expand their studies and advance human health.
Formalin-fixed paraffin-embedded (FFPE) tissue samples have proven vital to advancing cancer research. FFPE samples allow for a fuller analysis of precious samples, including investigation of important biomarkers, such as hotspot mutations, CNVs, gene fusions, and indels.
Synergetic biomarkers: Fueling advancements in cancer research
Oncology biomarker applications have helped transform our understanding and approach in the study of cancer. Explore this evolving field through the lens of these biomarkers and the advancements enabling breakthrough discoveries.
Download eBook
cfDNA isolation kits
Automated or manual extraction of high-quality enriched circulating cell-free DNA (cfDNA) from serum or plasma suitable for applications including real-time PCR, digital PCR, and next-generation sequencing.
Explore: MagMAX Cell-Free DNA Isolation Kit
Automated or manual extraction of high-quality DNA and RNA and miRNA from cell-free plasma, serum, or urine samples for use in applications such as Oncomine assays, next-generation sequencing (NGS), qPCR, and digital PCR.
Explore: MagMAX Cell-Free Total Nucleic Acid Kit
Cell-Free DNA Isolation Kit
Expand your possibilities—introducing additional sample purification capabilities. Isolate cell-free DNA from saliva and 10 mL of urine on KingFisher instruments.
Elevate your research
FFPE isolation kits
Automated and manual sequential isolations of DNA and RNA from the same formaldehyde- or paraformaldehyde-fixed paraffin-embedded (FFPE) tissue sample in separate eluates, both compatible with applications including real-time PCR and next-generation sequencing.
Explore: MagMAX FFPE DNA/RNA Ultra Kit
Combine MagMAX FFPE with AutoLys for a faster deparaffinization process
Speed up your deparaffinization process with the Applied Biosystems AutoLys system. Comprised of pretreated tubes and a rack system to reduce pipetting steps, the system eliminates the need for hazardous chemicals and minimizes loss of tissue sample.
Explore: AutoLys products
Liquid biopsy workflow
Use our cell-free total nucleic acid (cfTNA) kit or cfDNA kit with KingFisher instruments and Oncomine assays to optimize the liquid biopsy workflow from extraction and purification to analysis.
Liquid biopsy workflow for cfTNA and cfDNA extractions: sample collection, purification, library prep, sequencing, and analysis
Beads are better for cell free extractions
Beads bind the DNA more efficiently than glass fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of cellular particulates clogging these items during the extraction process. This clogging issue is of particular concern with protein-rich, large-volume samples such as plasma that are commonly used in cfDNA studies.
Leveraging Dynabeads magnetic beads
MagMAX kits leverage Dynabeads technology for sample extraction, one of the leading magnetic beads in the industry. The Dynabeads MyOne Silane beads contained in the MagMAX Cell-Free Isolation kits have an optimized silica-like surface chemistry with high specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads MyOne Silane ideal for the recovery of circulating cell-free nucleic acid targets present at low concentrations in cell-free body fluids or FFPE. Once captured, these fragments can then be eluted in small elution volumes of 15 µL to 50 µL and are ready for downstream applications.
Automate MagMAX Kits using KingFisher instruments
Four different Kingfisher purification instruments
Benefits of KingFisher instruments include
  • Various throughputs—Process 6–96 samples per run depending on the instrument model
  • Interchangeable formats— Process a wide range of input volumes
  • Protocol customization—Easily edit, modify, or create new protocols
  • Optimized reagents—Compatible with multiple magnetic-bead reagents
Explore: KingFisher instruments
Download: Validated software automation scripts for KingFisher purification instruments
Video Player is loading.
Current Time 0:00
Duration 5:40
Loaded: 0%
Stream Type LIVE
Remaining Time 5:40
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Video: How to purify circulating cell-free DNA (cfDNA)
    Learn how to perform manual and automated purification of circulating cell-free DNA from plasma samples. For more information, view the  User Guide for MagMAX Cell‑Free DNA.
    cfDNA extraction resources
    For Research Use Only. Not for use in diagnostic procedures.
    Stylesheet for Classic Wide Template adjustments
    Hero banner white gradient overlay