A complete CRISPR and TALEN gene editing toolkit
To help researchers in their quest to understand how the genome influences phenotype, we’ve developed a complete set of trusted solutions for every step in the gene editing workflow. Design your CRISPR-Cas9 system or TALEN constructs for precise cleavage, knock-in, and tagging, transfect them efficiently into cells, and validate the genotypic and phenotypic results. Our optimized, validated products, protocols, and tools work together to help eliminate the trial-and-error phase.
Because every lab is unique, we offer a variety of genome editing solutions to meet your needs. Whether you want results fast, seek full control over every design step, or need help engineering cells to your specific requirements, we have solutions that fit.
Download our genome editing resource guide
Video Player is loading.
Current Time 0:00
Duration 1:16
Loaded: 12.91%
Stream Type LIVE
Remaining Time 1:16
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    decorative image
    TrueGuide guide RNAs in as little as 5 days
    Innovation is at the core of what we do, and by optimizing our manufacturing procedures we're now able to ship custom and predesigned gRNAs in as little as 5 days to our North American customers.
    Gene editing tools and resources
    TrueDesign Genome Editor
    This free online tool enables scientists of all experience levels to easily design, select, and order reagents for precise gene editing experiments.
    Design your edit
    Complete CRISPR workflow
    We offer tools and solutions for every step in the CRISPR genome editing workflow. Our portfolio of genome editing products is built on 30 years of industry-leading innovation and can grow with your research needs.
    Explore the workflow
    High-performance Cas9 proteins
    Choose from our selection of Cas9 proteins, including our TrueCut Cas9 Protein v2 that delivers consistent high editing efficiency across gene targets and cell types, our high-fidelity Cas9 for minimization off-target effects, or our GMP-manufactured Cas9 for cell therapy applications.
    Choose your Cas9 protein
    CRISPR validated protocols
    Bypass the trial-and-error phase with step-by-step CRISPR protocols optimized for efficiency, viability, and reproducibility across a range of cell types and gene targets. 
    View protocols
    Genome Editing Learning Center
    Need to get started? Our free Learning Center includes educational articles, videos, webinars, and more covering the latest CRISPR and TALEN methods and best practices.
    Explore the Learning Center
    Genome editing workflow
    Genome editing experiments often progress through a basic standard workflow consisting of three design steps, a transfection step, and a validation step. This standard workflow is applicable to gene knockout, tagging, knock-in, and cell line engineering applications. Workflow steps are adaptable to fit each experiment. For example, knockout experiments and functional genomics screening may omit the knock-in step.
    The basic genome editing workflow includes steps for cleavage design, knock-in design, transfection, and validation
    Follow the links to learn more about our product offerings for each workflow step. In many cases, the TrueDesign Genome Editor can be used to design and order reagents for the complete editing experiment.
    Design and buildDeliverDetect and validate
    TrueGuide Synthetic gRNA
    TrueCut Cas9 Protein v2
    TrueTag Donor DNA Kit, GFP
    Neon Transfection System
    Attune NxT Flow Cytometer, blue
    Design tool
    CRISPR gRNAs
    CRISPR libraries
    Cas9 nucleases
    • Fluorescent or epitope tags
    • SNPs and indels
    • Selection markers
    • STOP codons
    Learn more
    • Transfection reagents
    • Electroporation instruments
    • Cell culture
    Learn more
    • Cleavage detection
    • Clonal isolation
    • Confirmation of genotype
    • Confirmation of phenotype
    Learn more
    TALEN mRNA pairs
    To see an example of this gene editing workflow in action, view the webinar on using the CRISPR-Cas9 system for CAR T cell knock-in.
    Choosing between CRISPR-Cas9 and TALEN gene editing platforms
    When designing your genome edit, you can choose between CRISPR or TALEN technologies.
    Although CRISPR-Cas9 gene editing requires a protospacer-adjacent motif (PAM) site in the vicinity of the desired break, this platform is generally viewed as simpler to design and implement. gRNAs can be made at low costs to target nearly any genomic sequence of interest, and expressing multiple gRNAs in cells allows for multiplexing of CRISPR edits.
    In contrast, TALEN usage is not dependent on the availability of a PAM site and can be used at any location in the genome. Closer proximity of the TALENs to the target insertion site can also lead to improved homology-directed repair (HDR). Although TALEN may have more specific and flexible capabilities, it is more time consuming and less cost-effective than CRISPR.
    Comparison between CRISPR vs TALEN gene editing technologies
    TechnologyCRISPR-Cas9TALEN
    End goalPermanent gene knockout or knock-inPermanent gene knockout or knock-in
    Key advantages
    • Excellent cleavage efficiency
    • Simple RNP assembly process
    • Multiplexing capability
    • Flexible design; no PAM requirement
    • Exceptional HDR efficiency due to closer proximity to insertion site
    • Includes rights under foundational TAL intellectual property
    Design restrictionsPAM site close to desired edit locus (NGG for S.pyogenes Cas9)None
    More informationLearn moreLearn more
    Explore toolsChoose Cas9 formatExplore TALEN choices
    Intended use of the products mentioned on this page varies. For specific intended use statements, please refer to the product label.
    Stylesheet for Classic Wide Template adjustments