SkanIt Microplate Reader Software
The all-in-one plate reader control and analysis software.
Thermo Scientific SkanIt Software streamlines data acquisition and analysis, helping ensure a seamless workflow that allows your research to advance with confidence.
Whether you require basic plate reading or more complex protocols with powerful analysis capabilities, the intuitive, efficient, and error-reducing SkanIt platform helps empower you to optimize your research, and offers superior control over the instrument settings for your Thermo Scientific microplate reader.
SkanIt Software for microplate readers
Discover the extensive features and benefits of this comprehensive plate reader control and analysis software.
Key benefits
SkanIt Software is easy to use
SkanIt Software is designed to conveniently serve multiuser environments. Operation is simple and straightforward, whether you need only basic plate reading capabilities or more complex protocols with powerful analysis. SkanIt Software offers flexible and versatile features for advanced applications. Given SkanIt Software is an open-license platform that authorizes unlimited installation privileges, analysis can be done outside of the lab while measurements are ongoing as you can easily transfer sessions between different computers.
SkanIt Software helps prevent errors
SkanIt Software incorporates various measures to help prevent errors like prompting users to rename experiments to avoid data mix-ups or loss. Measurement data from executed sessions are preserved unless the user intentionally deletes the file, allowing for experimentation and iterative design within the software. The specialized Drug Discovery Edition enhances data security by utilizing a database to include features that enable compliance with FDA 21 CFR Part 11, such as digital signatures, user login with Windows authentication, and comprehensive audit trails.
SkanIt Software increases productivity
SkanIt Software helps enhance productivity with its rapid processing capabilities and the option to analyze data on a computer separate from the main laboratory system. The software includes a royalty-free license, offers easily accessible data, provides comprehensive analysis tools for various common workflows, and supports automatic data import into custom-defined Microsoft Excel templates.
Features
Session management
SkanIt Software Research Edition saves data into program-specific files with the file extension .skax, which is automatically recognized by the Windows operating system. This makes opening of data files very fast and easy. Just double-click a .skax file in any folder to immediately launch SkanIt Software and open the data.
Virtual pipetting tool
This intuitive visual tool lets you quickly enter sample information in a manner that resembles reagent pipetting, making it easy to define the plate layout.
Measurement protocol
The measurement protocol setup is easy. Simply add the instrument actions to the session tree in the order they will be performed. The software offers flexible features for assays of different complexities.
Results display
Measurement data are continuously saved during the run, to help ensure that data are not lost due to unexpected interruptions such as power outages. The software has a clear display where data are shown in convenient result numbers, kinetic curves, spectral graphs, and 3D graphs. The software brings additional integrity to your research by saving important information with traceability. Temperatures, time stamps of actions, errors, and warnings are just a few of the prompts you will receive. With the optional integrated gas module, the log will also report the CO₂ and O₂ concentrations.
Data processing tools
Easily analyze your data directly in SkanIt Software. Select from a variety of data processing tools:
  • Average, SD, and %CV from sample replicates 
  • Curve fitting for typical biological responses, including parallel line analysis, enzyme kinetics (Km and Vmax), Z-factor, and many more 
  • Back-calculation of samples and standards for QC or assay verification using curve fitting
  • Classification for dividing samples into categories 
  • Normalization of data to your selected reference sample (e.g., for calculation of cell viability percentages) 
  • Calculations for reduction of kinetic or spectral data 
  • Merging multiple measurements into a single kinetic data set for better data analysis 
  • Advanced calculation tool for creating custom formulas
SkanIt Cloud Library
The SkanIt Cloud Library is a collection of verified sessions that anyone can use as examples, training materials, or templates to create assay protocols. The SkanIt Cloud Library sessions all have complete calculation protocols, with real assay data using samples and reagents. With the easy-to-use search feature, you can enter any keyword to find the protocol you need.
Exporting results 
SkanIt Software presents easily accessible, comprehensive reports of both measured and calculated data. Users can also select the way the data are organized and sorted—export the results report in .xlsx, .pdf, .xml, or .txt file format. Additionally, using the “auto loading” feature, you can load measurement data automatically into a predefined Excel file for further calculations.
Highlights of SkanIt Software
  • Install open-license software on multiple computers 
  • Choose from Research or Drug Discovery Edition with features for 21 CFR Part 11 compliance
  • Access user interface languages in English, French, German, Spanish, Portuguese, Italian, Russian, Japanese, and Chinese 
  • Rely on robotic automation interface 
  • Search the SkanIt Cloud Library featuring ready-to-use protocols that include calculations
  • Easily set up and navigate measurement sessions 
  • Define plate layouts using the Virtual Pipetting Tool 
  • Conduct single- or multiple-wavelength measurements, as well as multi-technology measurements
  • Perform endpoint, kinetic, spectral scanning, bottom-reading with multipoint option, and kinetic-spectral measurements 
  • Use built-in calculations for fast, accurate data analysis, including: 
    • Parallel line analysis 
    • Enzyme kinetic analysis of Km and Vmax 
    • Z-factor analysis 
    • Linear and logistic regression analysis 
  • Data transferable to a Microsoft™ Excel™ file with a single click 
  • Export data manually or automatically to .xlsx, .pdf, .xml, and .txt file formats 
  • Automatically load measurement data into a customizable Excel templates 
Sample images
Screenshot of the Virtual Pipetting Tool of SkanIt Software that helps visually define plate layout with blanks, standards, controls and unknowns. Innovative Virtual Pipetting Tool assists in defining samples within the plate layout.
Screenshot of session tree cascade The logical structure of the session tree is the main interface of SkanIt Software for Thermo Scientific microplate readers.
NA-Star assay for metabolites and analytes plotted in parallel line assay in SkanIt Softwar Relative potency calculation of two common antiviral drugs using Parallel Line Assay.
Beta-gal Km and Vmax calculated in SkanIt Software from β-galactosidase assay SkanIt software Enzyme Kinetics calculation tool calculates enzyme kinetic parameters Km and Vmax from the kinetic measurement data set.
PC requirements