Search Thermo Fisher Scientific
Discover a range of complimentary products for your microplate reader requirements, such as advanced microplate washers, high-precision multidrop dispensers, and efficient multichannel pipettes. Also explore a broad range of microplate assays by application and instrument compatibility.
Our reliable and easy-to-use Thermo Scientific Wellwash microplate strip washers provide secure washing performance for routine and research ELISA applications. Wellwash washers feature a large color display with graphical user interface, local language versions, and non-pressurized bottles for optimal ease-of-use, convenience, and safety.
Wash head | |
---|---|
1x8, 1x12 |
two 1x8, two 1x12, 2x8 cell wash head |
Wash bottles | |
1 bottle (2 litre) |
3 bottles (two 4 litre and one 2 litre) |
Plate formats | |
96 |
96, 384 |
Cell washing | |
No |
Yes |
Robot compatibility | |
No |
Yes |
Engineered to meet the diverse demands of liquid dispensing, Thermo Scientific Multidrop reagent dispensers are user-friendly and compact to fit easily on the bench with an intuitive user interface for effortless setup and programming. Achieve precise and reproducible plate filling in seconds, enabling faster test runs and enhanced productivity.
![]() Multidrop Combi+ |
![]() Multidrop Combi nL |
||
User interface |
On-board interface or FILLit PC software |
On-board interface or FILLit PC software |
On-board interface or FILLit PC software |
---|---|---|---|
Volume |
0.5–2500 µL | 0.5–2500 µL | 50 nL–50 µL |
Volume increments | Small tube dispensing cassette: 0.5–50 μL, in 0.5 μL increments Standard tube dispensing cassette: 5– 2,000 µL in 5 µL increments |
Small tube dispensing cassette: 0.5–50 μL, in 0.5 μL increments Standard tube dispensing cassette: 5– 2,000 µL in 5 µL increments |
1 nL increments 50–999 nL, 10 nL increments 1.00–9.99 µL, 100 nL increments 10.0–50.0 µL, FILLit Software 1 nL increments |
Dispense volume adjustability | Per column | Per column | Per well |
Dead volume | < 1 mL volume range: 0.5–50 µL <7 mL volume range: >50 µL (it is possible to backflush the reagent into original reservoir to avoid loss of expensive reagent) |
< 1 mL volume range: 0.5–50 µL <7 mL volume range: >50 µL (it is possible to backflush the reagent into original reservoir to avoid loss of expensive reagent) |
<1.2 mL |
Plate formats |
9- to 1,536-well plates: including 96- ad 384-deep well and tubes in rack format (plate height: 5–50 mm) | 9- to 1,536-well plates: including 96- ad 384-deep well and tubes in rack format (plate height: 5–50 mm) | 96-1536 well plates; including 96, 384- tubes in rack format |
Plate height adjustment |
Automatic | Automatic | Automatic |
Automation/robot compatible |
Yes | Yes | Yes |
Dispensing Technology |
Peristaltic pump | Peristaltic pump | Pressurized valve system |
Dimensions |
14.0 x 8.6 x 12.9 in. (35.6 x 21.8 x 32.8 cm) |
14.0 x 8.6 x 12.9 in. (35.6 x 21.8 x 32.8 cm) |
14 x 8.6 x 14.8 in 35.5 x 22 x 37.5 cm |
Weight | 20.1 lb/9.1K | 20.1 lb/9.1K | 21.2 lb/9.6 kg |
*Multidrop Combi SMART+ uses RFID to track cassette usage
Thermo Scientific offers a variety of multichannel pipettes for all applications. From the proven performance of our Finnpipette and Novus lines to the innovative ClipTip and ClipTip Equalizer pipettes, we are a leader in pipette technology and focused on providing worry-free performance with comfortable operation every day. For enhanced accuracy and precision, explore our range of manual, electronic, single-channel, and multichannel pipettes.
For Laboratory Use. It is the customer’s responsibility to ensure that the performance of the product is suitable for customers’ specific uses or applications.