Quick, safe and efficient evaporation 
SpeedVac systems use the state-of-the-art centrifugation, vacuum and heat technologies for removing solvents and concentrating samples while maintaining sample integrity. A wide and diverse product range is able to provide solutions for a wide variety of solvent applications ranging from the traditional drying down of DNA preparations in water and methanol, to the more complex and aggressive applications presented by the combinatorical chemistry and drug discovery sectors of the pharmaceutical industry.
We also offer simple prepackaged Speedvac Vacuum Concentrator Modular Kits for popular applications and a wide array of SpeedVac rotors.
Choose your individual SpeedVac Vacuum Concentrator
Choose from the individual SpeedVac Vacuum Concentrators below for protein, enzyme, and HPLC fraction applications
SPD120
SPD210
SPD130DLX
Programmability
333
Preset programs by solvent type333
User defined programs300
Capacity120 x 1.5mL tubes
2 deep well plates
6 shallow well plates
(and other tube & vial formats)
200 x 1.5mL tubes
4 deep well plates
12 shallow well plates
(and other tube & vial formats)
120 x 1.5mL tubes
2 deep well plates
6 shallow well plates
(and other tube & vial formats)
Type of lidGlassGlassRadiant Glass
Vacuum ramp / vacuum gaugeNoNoNo
Solvent typesWater, alcohol, methanol, acetonitrile, non-aggressive solventsWater, alcohol, methanol, acetonitrile, non-aggressive solventsWater, alcohol, methanol, acetonitrile, aggressive low boiling point solvents  
Order now
Choose from the individual SpeedVac Vacuum Concentrators below for organic solvents and drug discovery applications
SPD140DDA
SPD300DDA
Programmability
1212
Preset programs by solvent type44
User defined programs88
Capacity120 x 1.5mL
2 deep well plates
6 shallow well plates
(other tube & vial formats)
200 x 1.5mL
8 deep well plates
20 shallow well plates
(other tube & vial formats)
Type of lidHeated GlassHeated Glass
Vacuum ramp / vacuum gaugeYesYes
Solvent typesWater, low & high boiling, more aggressive solvents (DMSO, TFA), HPLCWater, low & high boiling, more aggressive solvents (DMSO, TFA), acids, bases, HPLC
 With PRO rotor systemWithout rotor
Choose from the individual SpeedVac Vacuum Concentrators below for protein, enzyme, and HPLC fraction applications
SPD120
SPD210
SPD130DLX
Programmability
333
Preset programs by solvent type333
User defined programs300
Capacity120 x 1.5mL tubes
2 deep well plates
6 shallow well plates
(and other tube & vial formats)
200 x 1.5mL tubes
4 deep well plates
12 shallow well plates
(and other tube & vial formats)
120 x 1.5mL tubes
2 deep well plates
6 shallow well plates
(and other tube & vial formats)
Type of lidGlassGlassRadiant Glass
Vacuum ramp / vacuum gaugeNoNoNo
Solvent typesWater, alcohol, methanol, acetonitrile, non-aggressive solventsWater, alcohol, methanol, acetonitrile, non-aggressive solventsWater, alcohol, methanol, acetonitrile, aggressive low boiling point solvents  
Order now
Choose from the individual SpeedVac Vacuum Concentrators below for organic solvents and drug discovery applications
SPD140DDA
SPD300DDA
Programmability
1212
Preset programs by solvent type44
User defined programs88
Capacity120 x 1.5mL
2 deep well plates
6 shallow well plates
(other tube & vial formats)
200 x 1.5mL
8 deep well plates
20 shallow well plates
(other tube & vial formats)
Type of lidHeated GlassHeated Glass
Vacuum ramp / vacuum gaugeYesYes
Solvent typesWater, low & high boiling, more aggressive solvents (DMSO, TFA), HPLCWater, low & high boiling, more aggressive solvents (DMSO, TFA), acids, bases, HPLC
 With PRO rotor systemWithout rotor
Smart Note: Savant SpeedVac System
When the quality of your results hinges on the quality of synthesized oligos
As oligos are being aggressively leveraged in research and clinical applications such as detecting, characterizing, and developing therapeutics and vaccines, the quality of synthesized oligos is critical. The Savant SpeedVac system enhances speed and reliability so you can have complete confidence in the quality of your oligos as well as the applications they serve.
Download Brochures
Features of SpeedVac Vacuum Concentrators
Focused on ease of use
Focused on ease of use
  • User interface allows use of 4 preset programs and 8 user defined programs to set heat, run time, ramp rate and vacuum level for efficient operation.
  • The heavy-duty universal rotor is optimized for efficient large-sample processing while accommodating various sample handling formats, such as microcentrifuge tubes, microplates, and 250mL flasks
Samples stay safe for reliable evaporation
Samples stay safe for reliable evaporation
  • Standard glass lid locks securely to help ensure safety for samples and users. Hands free operation when loading or unloading the rotor with the lid stay mechanism. An audible alarm alerts the user when the operation is finished.
Reliable data collection
Reliable data collection
  • Ability to download live run data from a USB port enhances data integrity.
Support
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.