SpeedVac kits
SpeedVac Kits for Every Lab
Thermo Scientific offers users a choice to purchase either an all in one fully integrated SpeedVac or to build a SpeedVac™ system to adapt for different solvents type and applications needs.
The fully integrated SpeedVac kits contain all components integrated into one compact system. Take the system out of the box, plug it in and it is ready operation.
Fully Integrated systems include the DNA130, SPD1030 and SPD2030 models.
For more aggressive high boiling solvents, use a modular system. Additionally, modular systems allow you the flexibility to replace the vacuum concentrator, add a secondary cold trap or use a different vacuum pump depending on the application needs.
Modular kits include:  SPD120, SPD130, SPD140, SPD210 and SPD300DDA.
Smart Note: Savant SpeedVac System
When the quality of your results hinges on the quality of synthesized oligos
As oligos are being aggressively leveraged in research and clinical applications such as detecting, characterizing, and developing therapeutics and vaccines, the quality of synthesized oligos is critical. The Savant SpeedVac system enhances speed and reliability so you can have complete confidence in the quality of your oligos as well as the applications they serve.
Download Brochures
Support
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.