Protein Gel welcome packs are cost-saving product combinations that contain all you need for outstanding protein separation. They include mini and midi format gels, buffers and reagents, as well as our Mini or Midi gel tank, with or without blot modules.
Protein gel welcome packs include:
  • Mini or Midi gel tank
  • 2 gel boxes
  • Running buffer
  • Sample buffer
  • Reducing agent
  • Prestained Ladder
  • Mini blot modules options available
  • WedgeWell format gel options available
Don’t need a tank? See our Protein Gel Resupply Packs to restock on protein gels and gel electrophoresis reagents.
Gel chemistryBolt Bis-Tris PlusNuPAGE Bis-TrisNovex Tris-GlycineNuPAGE Tris-AcetateNovex Tricine
Molecular Weight rangeBroad MW (6–250 kDa)Broad MW (6–250 kDa)Broad MW (6–250 kDa)High MW (40–500 kDa)Low MW (2.5–40 kDa)
Gel formatMiniMidi
Mini
Midi
Mini
Midi
Mini
Mini
WedgeWell format MiniMidiMidi and miniMidi and mini
Reduced protein degradationXX X 
Fast run timesXX   
Extended shelf lifeXXX  
Midi gel welcome pack options
Midi, wider format, gels support higher sample throughput needs by allowing the separation of up to 26 samples per gel, saving time and reagents and helping improve experimental reproducibility.
High loading capacity meets high sample throughput
WedgeWell format Midi protein gels feature wedge-shaped wells that double the sample loading capacity. WedgeWell format Midi gel welcome packs include a SureLock Tandem Tank and offer:
Wedgewell icon
WedgeWell format wells
—load up to 100 µL of samples
Protein gel icon
Wide format gels
—run up to 26 samples per gel
pipette icon
Easy loading
—no specialized pipette tips needed
Ordering Information
Catalog # Name Size Price (USD) Qty
STM4009 Novex™ Midi Tris-Glycine Plus Welcome Pack, 4 to 20%, 20-well, 1 Welcome Pk. 1 pack
970.00
STM4008 Novex™ Midi Tris-Glycine Plus Welcome Pack, 4 to 20%, 26-well, 1 Welcome Pk. 1 pack
968.00
STM5009 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 4 to 20%, 26-well, 1 Welcome Pk. 1 pack
978.00
STM5010 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 4 to 12%, 20-well, 1 Welcome Pk. 1 pack
978.00
STM5011 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 4 to 12%, 26-well, 1 Welcome Pk. 1 pack
978.00
STM4010 Novex™ Midi Tris-Glycine Plus Welcome Pack, 4 to 12%, 20-well, 1 Welcome Pk. 1 pack
968.00
STM5014 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 12%, 20-well, 1 Welcome Pk. 1 pack
978.00
STM4012 Novex™ Midi Tris-Glycine Plus Welcome Pack, 10%, 26-well, 1 Welcome Pk. 1 pack
972.00
STM5015 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 12%, 26-well, 1 Welcome Pk. 1 pack
978.00
STM4011 Novex™ Midi Tris-Glycine Plus Welcome Pack, 4 to 12%, 26-well, 1 Welcome Pk. 1 pack
968.00
STM5012 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 10%, 20-well, 1 Welcome Pk. 1 pack
978.00
STM5013 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 10%, 26-well, 1 Welcome Pk. 1 pack
978.00
STM4013 Novex™ Midi Tris-Glycine Plus Welcome Pack, 10%, 20-well, 1 Welcome Pk. 1 pack
966.00
STM5008 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 4 to 20%, 20-well, 1 Welcome Pk. 1 pack
978.00
Catalog # Name Size Price (USD) Qty
STM4009 Novex™ Midi Tris-Glycine Plus Welcome Pack, 4 to 20%, 20-well, 1 Welcome Pk. 1 pack
970.00
STM4008 Novex™ Midi Tris-Glycine Plus Welcome Pack, 4 to 20%, 26-well, 1 Welcome Pk. 1 pack
968.00
STM5009 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 4 to 20%, 26-well, 1 Welcome Pk. 1 pack
978.00
STM5010 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 4 to 12%, 20-well, 1 Welcome Pk. 1 pack
978.00
STM5011 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 4 to 12%, 26-well, 1 Welcome Pk. 1 pack
978.00
STM4010 Novex™ Midi Tris-Glycine Plus Welcome Pack, 4 to 12%, 20-well, 1 Welcome Pk. 1 pack
968.00
STM5014 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 12%, 20-well, 1 Welcome Pk. 1 pack
978.00
STM4012 Novex™ Midi Tris-Glycine Plus Welcome Pack, 10%, 26-well, 1 Welcome Pk. 1 pack
972.00
STM5015 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 12%, 26-well, 1 Welcome Pk. 1 pack
978.00
STM4011 Novex™ Midi Tris-Glycine Plus Welcome Pack, 4 to 12%, 26-well, 1 Welcome Pk. 1 pack
968.00
STM5012 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 10%, 20-well, 1 Welcome Pk. 1 pack
978.00
STM5013 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 10%, 26-well, 1 Welcome Pk. 1 pack
978.00
STM4013 Novex™ Midi Tris-Glycine Plus Welcome Pack, 10%, 20-well, 1 Welcome Pk. 1 pack
966.00
STM5008 Novex™ WedgeWell™ Midi Tris-Glycine Plus Welcome Pack, 4 to 20%, 20-well, 1 Welcome Pk. 1 pack
978.00
Mini gel welcome pack options
Four gel chemistries for tailored protein separations.
Bis-Tris and Tris-glycine are excellent for broad molecular weight range separation, and both feature a long shelf-life. The neutral pH of the Bis-Tris and the Bis-Tris Plus gel chemistries helps preserve protein integrity and offer fast run times.
The Invitrogen Novex Tris-Glycine Gels offer maximum protein separation and crisp, straight bands. NuPAGE Tris-Acetate Gels are recommended for the separation of high molecular weight proteins up to 500 kDa. Novex Tricine Gels are designed for the separation of low molecular weight proteins and peptides.
Wedgewell icon
WedgeWell format wells
—load up to 100 µL of samples
Rocket icon
Run your gel in just 20 minutes—
Bolt Bis-Tris Plus gels
Target icon
Gel chemistry tailored to your target protein
Gel tank, transfer modules, box of precast gels, bottles of buffers and reagents for protein electrophoresis.
Catalog # Name Size Price (USD) Qty
NW0012A Bolt™ Bis-Tris Welcome Pack, 12%, 10-well, 1 Welcome Pk. kit Each
690.00
NW0412G Bolt™ Bis-Tris Plus Welcome Pack and Mini Blot Modules, 4-12%, 10-well, 1 kit Each
1,386.00
NW0412H Bolt™ Bis-Tris Plus Welcome Pack and Mini Blot Modules, 4-12%, 12-well, 1 kit Each
1,386.00
NW0412I Bolt™ Bis-Tris Plus Welcome Pack and Mini Blot Modules, 4-12%, 15-well, 1 kit Each
1,386.00
NW0008C Bolt™ Bis-Tris Welcome Pack, 8%, 15-well, 1 Welcome Pk. kit Each
690.00
NW0412PEX Bolt™ Mini Bis-Tris Welcome Pack for Protein Expression (4–12%, 12 well) Each
782.00
NW0008A Bolt™ Bis-Tris Welcome Pack, 8%, 10-well, 1 Welcome Pk. kit Each
690.00
NW0008B Bolt™ Bis-Tris Welcome Pack, 8%, 12-well, 1 Welcome Pk. kit Each
690.00
NW0412A Bolt™ Welcome Pack A (4-12%, 10-well), 1 Welcome Pk. kit Each
658.65
Online Exclusive
690.00
Save 31.35 (5%)
NW0412B Bolt™ Welcome Pack B (4-12%, 15-well), 1 Welcome Pk. kit Each
658.65
Online Exclusive
690.00
Save 31.35 (5%)
NW0010B Bolt™ Bis-Tris Welcome Pack, 10%, 12-well, 1 Welcome Pk. kit Each
656.65
Online Exclusive
690.00
Save 33.35 (5%)
NW0412C Bolt™ Bis-Tris Welcome Pack, 4-12%, 12-well, 1 Welcome Pk. kit Each
658.65
Online Exclusive
690.00
Save 31.35 (5%)
NW0010C Bolt™ Bis-Tris Welcome Pack, 10%, 15-well, 1 Welcome Pk. kit Each
656.65
Online Exclusive
690.00
Save 33.35 (5%)
NW0012B Bolt™ Bis-Tris Welcome Pack, 12%, 12-well, 1 Welcome Pk. kit Each
690.00
NW0012C Bolt™ Bis-Tris Welcome Pack, 12%, 15-well, 1 Welcome Pk. kit Each
690.00
NW0010A Bolt™ Bis-Tris Welcome Pack, 10%, 10-well, 1 Welcome Pk. kit Each
656.65
Online Exclusive
690.00
Save 33.35 (5%)
Bolt Bis Tris- Plus
Catalog # Name Size Price (USD) Qty
NW0412F Bolt™ Bis-Tris Plus XCell SureLock™ Mini-Cell Welcome Pack, 4-12%, 15-well Each 1
758.65
Online Exclusive
820.00
Save 61.35 (7%)
NW0412D Bolt™ Bis-Tris Plus XCell SureLock™ Mini-Cell Welcome Pack, 4-12%, 10-well Each 1
758.65
Online Exclusive
820.00
Save 61.35 (7%)
NW0412E Bolt™ Bis-Tris Plus XCell SureLock™ Mini-Cell Welcome Pack, 4-12%, 12-well Each 1
758.65
Online Exclusive
820.00
Save 61.35 (7%)
Catalog # Name Size Price (USD) Qty
NW0012A Bolt™ Bis-Tris Welcome Pack, 12%, 10-well, 1 Welcome Pk. kit Each
690.00
NW0412G Bolt™ Bis-Tris Plus Welcome Pack and Mini Blot Modules, 4-12%, 10-well, 1 kit Each
1,386.00
NW0412H Bolt™ Bis-Tris Plus Welcome Pack and Mini Blot Modules, 4-12%, 12-well, 1 kit Each
1,386.00
NW0412I Bolt™ Bis-Tris Plus Welcome Pack and Mini Blot Modules, 4-12%, 15-well, 1 kit Each
1,386.00
NW0008C Bolt™ Bis-Tris Welcome Pack, 8%, 15-well, 1 Welcome Pk. kit Each
690.00
NW0412PEX Bolt™ Mini Bis-Tris Welcome Pack for Protein Expression (4–12%, 12 well) Each
782.00
NW0008A Bolt™ Bis-Tris Welcome Pack, 8%, 10-well, 1 Welcome Pk. kit Each
690.00
NW0008B Bolt™ Bis-Tris Welcome Pack, 8%, 12-well, 1 Welcome Pk. kit Each
690.00
NW0412A Bolt™ Welcome Pack A (4-12%, 10-well), 1 Welcome Pk. kit Each
658.65
Online Exclusive
690.00
Save 31.35 (5%)
NW0412B Bolt™ Welcome Pack B (4-12%, 15-well), 1 Welcome Pk. kit Each
658.65
Online Exclusive
690.00
Save 31.35 (5%)
NW0010B Bolt™ Bis-Tris Welcome Pack, 10%, 12-well, 1 Welcome Pk. kit Each
656.65
Online Exclusive
690.00
Save 33.35 (5%)
NW0412C Bolt™ Bis-Tris Welcome Pack, 4-12%, 12-well, 1 Welcome Pk. kit Each
658.65
Online Exclusive
690.00
Save 31.35 (5%)
NW0010C Bolt™ Bis-Tris Welcome Pack, 10%, 15-well, 1 Welcome Pk. kit Each
656.65
Online Exclusive
690.00
Save 33.35 (5%)
NW0012B Bolt™ Bis-Tris Welcome Pack, 12%, 12-well, 1 Welcome Pk. kit Each
690.00
NW0012C Bolt™ Bis-Tris Welcome Pack, 12%, 15-well, 1 Welcome Pk. kit Each
690.00
NW0010A Bolt™ Bis-Tris Welcome Pack, 10%, 10-well, 1 Welcome Pk. kit Each
656.65
Online Exclusive
690.00
Save 33.35 (5%)
Bolt Bis Tris- Plus
Catalog # Name Size Price (USD) Qty
NW0412F Bolt™ Bis-Tris Plus XCell SureLock™ Mini-Cell Welcome Pack, 4-12%, 15-well Each 1
758.65
Online Exclusive
820.00
Save 61.35 (7%)
NW0412D Bolt™ Bis-Tris Plus XCell SureLock™ Mini-Cell Welcome Pack, 4-12%, 10-well Each 1
758.65
Online Exclusive
820.00
Save 61.35 (7%)
NW0412E Bolt™ Bis-Tris Plus XCell SureLock™ Mini-Cell Welcome Pack, 4-12%, 12-well Each 1
758.65
Online Exclusive
820.00
Save 61.35 (7%)
Find an excellent protein gel chemistry for your research needs
Decision tree for choosing a protein gel chemistry welcome pack for the right application and research needs
 Click image to enlarge
The choice of whether to use one chemistry or another depends on the abundance of the target protein, the size of the protein, and the downstream application.
Protein Gel Selection Guide
Protein Gel Selection Guide
Find an excellent mini or midi gel for your protein electrophoresis experiment.
Gel Selection Chart
Protein Gel Conversion Tool
Find an exceptional Invitrogen mini or midi gel to replace your current precast gel from another supplier.
Protein Gel Options Brochure
Gel Migration Chart
Download the Protein Gel Guide for a printable gel migration chart.
Still unsure which precast gel works best for your research needs? Download the Gel Selection Chart
Protein Gel Performance Guarantee
Invitrogen precast protein gels are designed and tested to deliver excellent performance, reliability, reproducibility, and consistency of quality. We guarantee it. Download our Certificate of Analysis for more details. No other supplier conducts such stringent quality testing.
Protein Gels Performance Guarantee
Protein Gels Performance Guarantee
We stand behind the performance of our protein gels.
Learn more
Hero banner dark gradient overlay
Stylesheet for Classic Wide Template adjustments
For Research Use Only. Not for use in diagnostic procedures.