Long-wavelength Qdot® and Alexa Fluor® conjugates for sensitive and quantitative detection
(See a list of the products featured in this article.)
The breakneck pace of cell biology research demands more sensitive and quantitative protein analysis methods, capable of dissecting increasingly complex protein expression and interaction patterns. Western blot detection methods have evolved from radioisotope-based detection and colorimetric methods to those based on enhanced chemiluminescence (ECL), which are at least 10 times more sensitive than colorimetric assays. With recent advances in Qdot® probe technology, near-IR–emitting fluorescent dyes, and fluorescence imaging platforms, researchers are now able to obtain detection sensitivities from fluorescence-based detection comparable with those obtained from ECL (Figure 1). Furthermore, unlike ECL techniques, fluorescence-based detection methods provide the capability to detect multiple antigens on a single blot without stripping and reprobing.
 Figure 1. Comparison of enhanced chemiluminescence (ECL)– and Qdot® fluorescence–based western blot protein detection. Serial dilutions (750–3 ng protein) of GAPDH were run on NuPAGE® Novex® 4–12% Bis-Tris precast gels and transferred to iBlot® nitrocellulose membranes using the iBlot® Gel Transfer Device. The membranes were then probed with mouse anti-GAPDH antibodies followed by either ECL detection using a horseradish peroxidase goat anti–mouse IgG conjugate or fluorescence detection using Qdot® 655 goat anti–mouse IgG conjugate. Images were collected using the Fujifilm® LAS-4000 gel imager. Partial blot images show GAPDH detection, and the graph shows the detection sensitivities of the two techniques. AU = arbitrary units.
Advantages of fluorescent probes for western blots
Qdot® and Alexa Fluor® secondary antibody conjugates are revolutionizing western blot detection. When paired with a fluorescence imager, these detection reagents provide sensitive and quantitative analysis of proteins blotted or spotted onto membranes. Importantly, probes with long-wavelength (red to near-IR) emission allow signal collection beyond the typical autofluorescence emitted by standard nitrocellulose and PVDF membranes, resulting in low background fluorescence and superior signal-to-noise ratios.
Tremendous flexibility in experimental design is afforded by the availability of several different long-wavelength Qdot® and Alexa Fluor® fluorophores, facilitating the detection of multiple proteins on a single western blot (Figures 2 and 3). A multicolor western blot saves time and sample by eliminating the need to produce duplicate gels and blots or to strip and reprocess a single blot with a different antibody probe. Moreover, data analysis is greatly simplified because you can directly compare signals from protein bands that have been exposed to identical electrophoresis and transfer conditions.
Multiplex westerns with Qdot® antibody conjugates
Qdot® secondary antibody conjugates provide a powerful alternative to traditional methods for western blot detection [1–3]. Furthermore, our new WesternDot™ antibodies utilize Qdot® nanocrystals enhanced with VIVID® technology, making them brighter than the original Qdot® reagents. Additionally, these WesternDot™ conjugates have reduced intensity differences between the different colors, simplifying multiplex applications. We offer the following WesternDot™ secondary antibody conjugates:
  • WesternDot™ 585 anti–mouse IgG (W10800, W10803), anti–rabbit (W10801, W10804), and anti–goat IgG (W10802) antibody conjugates
  • WesternDot™ 625 anti–mouse IgG (W10805, W10808), anti–rabbit (W10806, W10809), and anti–goat IgG (W10807) antibody conjugates (available by phone order only until December 15, when they will also be available online)
  • WesternDot™ 655 anti–mouse IgG (W10810, W10813), anti–rabbit (W10811, W10814), and anti–goat IgG antibody conjugates (W10812)
  • WesternDot™ 800 anti–mouse IgG (W10815) and anti–rabbit IgG antibody (W10816) conjugates
Qdot® nanocrystals are exceptionally bright and photostable fluorophores that can be excited at any wavelength below their emission (though most efficiently by UV, violet, or deep-blue light). Qdot® fluorescence can be detected on many commercially available fluorescence imagers, including relatively inexpensive gel imagers equipped with a UV light box and appropriate emission filters—e.g., the E-Gel® Imager with the Qdot® 625 filter. More advanced fluorescence imagers, such as the Fujifilm® LAS-4000 gel imager, enable higher-order multiplexing and quantitation.
Using protein-specific primary antibodies and Qdot® secondary antibody conjugates, we have probed a single western blot for two proteins of interest (EGFR and phosphorylated EFGR), as well as a third protein (GAPDH) for normalizing signals (Figure 2). In this example, multiplex detection facilitates the normalization of fluorescent signals and allows us to distinguish proteins that co-migrate in a single gel lane, enabling the identification and quantitation of posttranslational modifications when specific primary antibodies for these modifications are available. This type of multicolor analysis not only reduces workflow significantly by minimizing the need for multiple blots, but also makes the quantitation of multiple proteins much more reliable.
Figure 2. Simultaneous detection of three proteins on a single blot using Qdot® secondary antibody conjugates.
A western blot containing serial dilutions (20–3 µg protein) of lysates from unstimulated (lanes 2–5) and hEGF-stimulated (lanes 6–9) A431 cells was probed with mouse anti-EGFR, rabbit anti–phospho-EGFR, and chicken anti-GAPDH antibodies, followed by (A) Qdot® 800 goat anti-mouse (pseudocolored blue), (B) Qdot® 605 goat anti-rabbit (pseudocolored red), and (C) Qdot® 655 goat anti-chicken (pseudocolored green) conjugates. (D) The merged image shows overlaid red and blue bands as purple. The blot contains MagicMark™ XP Western Protein Standard (lane 1) and was imaged using the Fujifilm® LAS-4000 gel imager.
Multiplex westerns with Alexa Fluor® antibody conjugates
The near-IR–emitting Alexa Fluor® secondary antibodies are also valuable immunodetection tools that can be imaged and quantitated using near-IR fluorescence scanners (Figures 3 and 4). Figure 4 shows the detection of EGFR and phosphorylated EGFR with Alexa Fluor® 790 and Alexa Fluor® 680 antibody conjugates, respectively. Because the two proteins co-migrate in the lysates from hEGF-stimulated cells, the overlaid red and green bands appear yellow in the merged image. Although not resolved by molecular weight, each labeled protein can be individually detected and quantitated by the fluorescence imager.
Figure 3. Western blot detection of AKT and GAPDH using Alexa Fluor® antibody conjugates.
A western blot containing serial dilutions of Jurkat cell lysate (lanes 2–5) was probed with mouse anti-GAPDH and rabbit anti-AKT antibodies followed by Alexa Fluor® 790 goat anti–mouse IgG (pseudocolored red) and Alexa Fluor® 680 goat anti–rabbit IgG (pseudocolored green) conjugates, and then imaged using the LI-COR® Odyssey® Imaging System. Lane 1: MagicMark™ XP Western Protein Standard.
Figure 4. Simultaneous detection of EGFR and phospho-EGFR using Alexa Fluor® antibody conjugates.
A western blot containing lysates from unstimulated (lanes 2, 3) and hEGF-stimulated (lanes 4, 5) A431 cells was probed with mouse anti-EGFR and rabbit anti–phospho-EGFR antibodies followed by Alexa Fluor® 790 anti–mouse IgG (pseudocolored red) and Alexa Fluor® 680 anti–rabbit IgG (pseudocolored green) conjugates, and imaged using the LI-COR® Odyssey® Imaging System. Lane 1: MagicMark™ XP Western Protein Standard.
An increasingly fluorescent future
As researchers migrate from ECL to multiplex fluorescence-based western detection, the number of fluorescent probes and imaging platforms continues to increase. We are developing and testing our reagents on the latest platforms so that beginners and experts alike can get the most out of their assays.
Article download
Download a hyperlink-enabled, printer-friendly version of this article.
Download now
Ordering information
Catalog # Name Size Price (USD) Qty
W10800 Donkey anti-Mouse IgG (H+L) Secondary Antibody, WesternDot™ 585 25 blots -
W10801 Donkey anti-Rabbit IgG (H+L) Secondary Antibody, WesternDot™ 585 25 blots -
W10802 Donkey anti-Goat IgG (H+L) Secondary Antibody, WesternDot™ 585 25 blots -
W10803 Goat anti-Mouse IgG (H+L) Secondary Antibody, WesternDot™ 585 25 blots -
W10804 Goat anti-Rabbit IgG (H+L) Secondary Antibody, WesternDot™ 585 25 blots -
W10805 Donkey anti-Mouse IgG (H+L) Secondary Antibody, WesternDot™ 625 25 blots -
W10806 Donkey anti-Rabbit IgG (H+L) Secondary Antibody, WesternDot™ 625 25 blots
288.00
W10807 Donkey anti-Goat IgG (H+L) Secondary Antibody, WesternDot™ 625 25 blots -
W10808 Goat anti-Mouse IgG (H+L) Secondary Antibody, WesternDot™ 625 25 blots
312.00
W10809 Goat anti-Rabbit IgG (H+L) Secondary Antibody, WesternDot™ 625 25 blots
315.00
W10810 Donkey anti-Mouse IgG (H+L) Secondary Antibody, WesternDot™ 655 25 blots -
W10811 Donkey anti-Rabbit IgG (H+L) Secondary Antibody, WesternDot™ 655 25 blots -
W10812 Donkey anti-Goat IgG (H+L) Secondary Antibody, WesternDot™ 655 25 blots -
W10813 Goat anti-Mouse IgG (H+L) Secondary Antibody, WesternDot™ 655 25 blots -
W10814 Goat anti-Rabbit IgG (H+L) Secondary Antibody, WesternDot™ 655 25 blots -
W10815 Goat anti-Mouse IgG (H+L) Secondary Antibody, WesternDot™ 800 25 blots -
W10816 Goat anti-Rabbit IgG (H+L) Secondary Antibody, WesternDot™ 800 25 blots -
Q11001MP F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, Qdot™ 605 200 µL
526.00
Q11401MP F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, Qdot™ 605 200 µL
526.00
A10195 F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, Qdot™ 625 100 µg -
A10194 F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, Qdot™ 625 100 µL
332.00
Q11061MP F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, Qdot™ 705 200 µL -
Q11461MP F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, Qdot™ 705 200 µL
390.65
Online Exclusive
520.00
Save 129.35 (25%)
A21057 Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 680 1 mg
284.00
A21058 Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 680 1 mg
349.00
A21076 Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 680 1 mg
284.00
A21109 Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 680 1 mg
341.00
A11375 Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 790 1 mg
284.00
A11357 Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 790 1 mg
341.00
A11367 Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 790 1 mg
290.00
A11369 Goat anti-Rabbit IgG (H+L) Highly Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 790 1 mg
341.00
For Research Use Only. Not for use in diagnostic procedures.