Harness immune checkpoints to combat tumors
(See a list of the products featured in this article.)
As critical members of the adaptive immune system, T cells are capable of mounting an efficient immune response against tumor cells. As T cells circulate in the body, they are constantly interrogating proteins on the surface of cells they encounter. If a cell is identified as “foreign”, the T cells will unleash an attack against the invading cell. Tumor cells, however, can evade detection by masking their surface with proteins of normal cells. These proteins—referred to as immune checkpoints—can affect immunoregulatory pathways by either boosting (co-stimulatory) or restricting (co-inhibitory) the immune responses of T cells [1,2]. So far, a great number of immune checkpoint proteins have been identified (Figure 1).
immunecheck-fig1
Figure 1. Multiple co-stimulatory and co-inhibitory receptor–ligand interactions between antigen-presenting cells (APCs) and T cells. T cell receptors (TCRs) detect antigens on the surface of APCs in the form of antigen-complexed major histocompatibility complexes (MHCs), and this antigen-specific recognition is necessary but not sufficient for an effective T cell response. For T cell activation or suppression, T cells must recognize their cognate antigens through TCRs and then respond to co-stimulatory (for activation) or co-inhibitory (for suppression) receptor–ligand interactions, examples of which are shown in this schematic. One important family of membrane-bound molecules that bind both co-stimulatory and co-inhibitory receptors is the B7-CD28 family shown in purple boxes; all of the B7 family members and their known ligands belong to the immunoglobulin superfamily. Another major category of signals arise from tumor necrosis factor (TNF) family members (shown in green boxes), which regulate the activation of T cells in response to cytokines.
Antibodies for dissecting the immune checkpoint pathways
Antibodies that target immune checkpoint pathways—such as PD-1/PD-L1 (Figure 2), CTLA4/CD80 and CD86, LAG3/MHC II, and TIM3/GAL9—have proven effective as immunotherapeutic agents in cancer treatment. For example, PD-1/PD-L1 is a co-inhibitory pathway that functions to restrict the immune system by constraining T cell anti-tumor activity. Studies with antibodies (e.g., nivolumab and pembrolizumab) that target the PD-1/PD-L1 pathway and suppress its co-inhibitory function have shown durable clinical responses in combating certain cancers, even in patients with advanced-stage cancer [1,2].
Recently, the co-inhibitory receptor TIGIT has been found to be expressed on the surface of a variety of lymphocytes, including effector and regulatory CD4+ T cells, follicular helper CD4+ T cells, effector CD8+ T cells, natural killer (NK) cells, and memory T cells (Figure 3). The expression of TIGIT is markedly enriched on tumor-infiltrated T cells [3]. In addition, the TIGIT+ regulatory T cells in tumor tissue co-express TIM3, and TIM3 and TIGIT work together to suppress anti-tumor immunity [3]. Similarly, therapeutic agonists of DR3, a co-stimulatory receptor that is expressed primarily in tissues enriched in lymphocytes (Figure 4), can also be used to stimulate regulatory T cell expansion, which can reduce inflammation and enable patients to produce an effective anti-tumor response [4]. These examples show that releasing the anti-tumor immune response by blocking co-inhibitory immune checkpoints or activating co-stimulatory immune checkpoints is a promising approach for triggering anti-tumor responses and mediating durable cancer regressions.
immunecheck-fig2a
immunecheck-fig2b
immunecheck-fig2c
immunecheck-fig2d
Figure 2. The blockade of immune checkpoint PD-1/PD-L1 in immunotherapy. This figure shows PD-1 and PD-L1 for illustrative purposes, although the concept likely applies to multiple immune checkpoints. (A) When PD-1 on the T cell surface binds its ligand PD-L1 on the tumor cell, the T cell becomes inactivated, leaving the tumor cell intact and growing. Blocking the activity of PD-1 or PD-L1 with inhibitors (e.g., antibodies) can prevent the interaction of PD-1 and PD-L1, enabling the T cell to stay active and launch an anti-tumor immune response to the tumor cell, releasing inflammatory signals such as IFN-gamma. (B) Immunohistochemical staining of human brain tissue was performed using an anti-PD1 (CD279) polyclonal antibody (25 μg/mL). (C, D) Unstimulated or PHA-stimulated normal human peripheral blood cells have been stained with PD-1 or PD-L1 antibody as follows, and viable cells in the lymphocyte gate were detected using Invitrogen eBioscience Fixable Viability Dye eFluor 520. PD-1 or PD-L1 antibodies include: (C)APC mouse IgG1 kappa isotype control (blue histogram) or APC anti–human CD279 (PD-1) antibody (purple histogram). (D)APC mouse IgG1 kappa isotype control (blue histogram) or APC anti–human CD274 (PD-L1, B7-H1) antibody (purple histogram).
ImmuneCheck_Fig3
Figure 3. Expression of TIGIT on CD4+ CD45RO+ T cells. Normal human peripheral blood cells were stained with APC anti–human CD4 antibody (clone RPA-T4, Cat. No. 17-0049-42), PerCP-eFluor 710 anti–human CD45RO antibody (clone UCHL1, Cat. No. 46-0457-42), and PE mouse IgG1 kappa isotype control (top, Cat. No. 12-4714-82) or PE anti–human TIGIT antibody (bottom, Cat. No. 12-9500-42). CD4+ cells in the lymphocyte gate were used for analysis.
ImmuneCheck_Fig4v3
Figure 4. Expression of DR3 in Ramos cells. Immunocytochemical fluorescence analysis was performed on fixed and permeabilized Ramos cells for detection of (A) endogenous DR3 (TNFRSF25) using Invitrogen anti-DR3 (TNFRSF25) ABfinity recombinant rabbit monoclonal antibody (2 μg/mL) in conjunction with Invitrogen Alexa Fluor 488 goat anti–rabbit IgG Superclonal secondary antibody (green, 1:2000 dilution), (B) nuclei using Invitrogen SlowFade Gold Antifade Mountant with DAPI (blue), and (C) cytoskeletal F-actin using rhodamine phalloidin (red, 1:300 dilution). (D) The composite image shows staining of DR3 (TNFRSF25), nuclei, and actin. To assess background fluorescence, control cells were stained similarly except that no primary antibody was used (data not shown). The images were captured at 60x magnification on a Nikon™ Eclipse™ Ti-U inverted microscope.
Check out our checkpoint antibodies
Despite recent breakthroughs in immunotherapy to treat melanoma of the skin, non-small cell lung cancer, kidney and bladder cancers, head and neck cancers, and Hodgkin lymphoma, the complex biology of the immune checkpoint pathways is still far from being understood. We offer a wide range of antibodies designed to study immune checkpoints (Table 1), as well as blocking antibodies, proteins, assays, and more. Our experienced custom service team can also save you time by developing antibodies to meet your specifications.
Table 1. Selected antibodies for flow cytometry, immunohistochemistry (IHC)/immunofluorescence (IF), and functional bioassays.
Immune checkpointAnti-human antibody (Cat. No.)Anti-mouse antibody (Cat. No.)
Flow cytometryIHC/IFFunctional bioassayFlow cytometryIHC/IFFunctional bioassay
B7-H3 (CD276)MA1-74441, 46-2769-42MA5-15693, MA1-7444116-5937-8516-5973-81 16-5973-81
B7-H4 (VTCN1)MA5-16845, 17-5949-42MA1-74439, MA1-7444016-5949-8212-5970-83, 12-5972-82 16-5972-81
B7-H5 (VISTA) PA5-52493 46-5919-82PA5-4703216-5919-85
BTLA (CD272)MA5-16843, 13-5979-80PA5-22248, MA1-7421216-5979-8212-5950-82, 17-5956-82 16-5950-82
CD2711-0279-42, 11-0271-8516-0272-85, PA5-2803616-0271-85MA5-17902, MA5-1790412-0271-8316-0272-85
CD2861-0289-42, MA1-10170MA1-10166, MA5-1700516-0281-86MA1-34061, 14-0281-86MA1-1017216-0289-85
CD30MA5-12632, 11-0309-42MA5-12632 12-0301-82MA1-9023416-0301-85
CD30L (CD153)MA5-23859PA5-47045MA5-23859A18356, 12-1531-8314-1531-8514-1531-85
CD40MA1-80926, 11-0409-4214-0409-82, 70012116-0409-85MA5-17853, 46-0401-8214-0401-8616-0402-86
CD40L (CD154)MA1-33895, 11-1548-42PA5-1348316-1548-82HMCD15401, 46-1541-82MA1-1010816-1541-85
CD70MA5-17727, 50-0709-42PA5-32701, MA5-17726 14-0701-85, MA1-81645 16-0701-85
CD80MA1-19215, 11-0809-42MA5-15512, MA1-9087216-0809-85MA1-70090, 11-0801-8614-0801-8516-0801-85
CD86MA1-81186, 12-0869-42MA1-12172, MA1-1029314-0869-8212-0861-83, 11-0862-8514-0862-8516-0861-85
CD96 (Tactile)46-0969-42, MA5-24280PA5-64286 MA5-24281, 12-0960-80 16-0960-82
CD112 (Nectin-2)MA1-35890, 17-1128-42PA5-29757    
CD134 (OX40)MA5-23591, 11-1347-42PA5-34516, 14-1347-82 12-1341-83, MA1-7008714-1341-85, MA5-1791616-1341-85
CD137 (4-1BB)MA5-13739, 11-1379-42MA5-13739PA5-47037PA5-47967, 25-1371-82 16-1371-85
CD137L (4-1BB L)PA5-47291  46-5901-82  
CD152 (CTLA-4)12-1528-42, 25-1529-42PA5-23967, PA5-4754716-1529-8261-1522-82, HMCD15201 16-1521-85
CD155 (PVR)MA5-13493, 46-1550-42MA5-13490 17-1551-82MA5-24315 
CD223 (LAG3)46-2239-42  MA5-24284, 11-2231-82 16-2231-85
CD226 (DNAM1) PA5-38444, PA5-31111 MA5-17990, 16-2261-85PA5-3639316-2261-85
CD252 (OX40L)PA5-24552PA5-34516 46-5905-82  
CD258 (LIGHT)46-2589-42, 17-2589-42     
CD273 (PD-L2)17-5888-42, MA5-16839PA5-2034416-5888-8212-5986-83, 11-9972-8514-5986-8516-5986-82
CD274 (PD-L1)46-5983-42, MA5-1684014-5983-8216-5983-8262-5982-80, 13-9971-8114-5982-8516-5982-85
CD278 (ICOS)11-9948-42, MA5-2368014-9948-8216-9948-8246-9940-82, 11-9942-8214-9949-8216-9942-85
CD279 (PD-1)12-9969-42, 12-2799-4214-2799-80, 14-9969-8216-9989-3811-9981-82, 25-9985-8214-9985-8516-9985-85
CD357 (GITR)48-5875-42, MA5-23854PA5-46810, PA5-47883PA5-47883MA5-17934, 11-5874-8216-5874-8316-5874-83
DR3 (TNFRSF25)702277, 12-6603-42PA1-30533, PA5-28293 711309, MA5-23838711309 
Galectin-950-9116-42PA5-32252, PA5-5096616-9116-85PA5-47503MA5-24369 
GITRL PA5-20161MA5-23852, PA5-47885 PA5-20161 
HVEMMA1-25958, 12-5969-80PA5-20236, PA5-26103 17-5962-82PA5-2023716-5962-85
ICOSL (B7RP1, B7-H2)MA1-17756, 12-5889-4213-5889-8216-5889-8250-5985-82, MA5-24270PA5-4716116-5985-85
IDOMA5-23595, 12-9477-42PA5-29819, PA5-12305 46-9473-82PA5-24598 
TIGIT46-9500-42 16-9500-8512-9501-82 16-9501-85
TIM347-3109-42 16-3109-8516-5871-85, MA5-179514-5871-8516-5871-85
TL1A46-7911-82  46-7911-82  
VSIG417-5757-42PA5-52018 17-5752-82  
Catalog # Name Size Price (USD) Qty
16-2261-85 CD226 (DNAM-1) Monoclonal Antibody (10E5), Functional Grade, eBioscience™ 500 µg
453.90
Special offer
Online exclusive
534.00
Save 80.10 (15%)
16-1371-85 CD137 (4-1BB) Monoclonal Antibody (17B5), Functional Grade, eBioscience™ 500 µg
315.65
Online Exclusive
420.00
Save 104.35 (25%)
16-0801-85 CD80 (B7-1) Monoclonal Antibody (16-10A1), Functional Grade, eBioscience™ 500 µg
399.00
12-1531-83 CD153 Monoclonal Antibody (RM153), PE, eBioscience™ 200 µg
370.00
PA529757 Nectin 2 Polyclonal Antibody 100 µL
498.00
16-1548-82 CD154 (CD40 Ligand) Monoclonal Antibody (24-31), Functional Grade, eBioscience™ 100 µg
298.35
Special offer
Online exclusive
351.00
Save 52.65 (15%)
16-0809-85 CD80 (B7-1) Monoclonal Antibody (2D10.4), Functional Grade, eBioscience™ 500 µg
396.10
Special offer
Online exclusive
466.00
Save 69.90 (15%)
16-0289-85 CD28 Monoclonal Antibody (CD28.2), Functional Grade, eBioscience™ 500 µg
365.50
Special offer
Online exclusive
430.00
Save 64.50 (15%)
14-0409-82 CD40 Monoclonal Antibody (5C3), eBioscience™ 100 µg
122.40
Special offer
Online exclusive
144.00
Save 21.60 (15%)
11-1548-42 CD154 (CD40 Ligand) Monoclonal Antibody (24-31), FITC, eBioscience™ 100 tests
294.95
Special offer
Online exclusive
347.00
Save 52.05 (15%)
MA181645 CD70 Monoclonal Antibody (FR70), FITC 500 µg -
MA517727 CD70 Monoclonal Antibody (BU69), FITC 100 µg
398.00
16-0701-85 CD70 Monoclonal Antibody (FR70), Functional Grade, eBioscience™ 500 µg
362.10
Special offer
Online exclusive
426.00
Save 63.90 (15%)
MA517726 CD70 Monoclonal Antibody (BU69), Biotin 100 µg
435.00
61-0289-42 CD28 Monoclonal Antibody (CD28.2), PE-eFluor™ 610, eBioscience™ 100 tests
323.85
Special offer
Online exclusive
381.00
Save 57.15 (15%)
25-1371-82 CD137 (4-1BB) Monoclonal Antibody (17B5), PE-Cyanine7, eBioscience™ 100 µg
364.65
Special offer
Online exclusive
429.00
Save 64.35 (15%)
14-0401-86 CD40 Monoclonal Antibody (1C10), eBioscience™ 1 mg
545.70
Special offer
Online exclusive
642.00
Save 96.30 (15%)
11-2231-82 CD223 (LAG-3) Monoclonal Antibody (eBioC9B7W (C9B7W)), FITC, eBioscience™ 100 µg
203.15
Special offer
Online exclusive
239.00
Save 35.85 (15%)
16-5919-85 VISTA Monoclonal Antibody (MH5A), Functional Grade, eBioscience™ 500 µg
562.70
Special offer
Online exclusive
662.00
Save 99.30 (15%)
11-0271-85 CD27 Monoclonal Antibody (LG.7F9), FITC, eBioscience™ 500 µg
317.05
Special offer
Online exclusive
373.00
Save 55.95 (15%)
PA528036 CD27 Polyclonal Antibody 100 µL
474.00
MA190234 CD30 Monoclonal Antibody (Ber-H2) 500 µL -
HMCD15401 CD154 (CD40 Ligand) Monoclonal Antibody (MR1), FITC 500 µg
590.00
11-0809-42 CD80 (B7-1) Monoclonal Antibody (2D10.4), FITC, eBioscience™ 100 tests
220.15
Special offer
Online exclusive
259.00
Save 38.85 (15%)
MA190872 CD80 Monoclonal Antibody (MAB104), FITC 50 tests -
MA112172 CD86 Monoclonal Antibody (B72-H2) 100 µg -
MA513490 CD155 Monoclonal Antibody (D171), Biotin 500 µL
522.00
46-1541-82 CD154 (CD40 Ligand) Monoclonal Antibody (MR1), PerCP-eFluor™ 710, eBioscience™ 100 µg
311.10
Special offer
Online exclusive
366.00
Save 54.90 (15%)
16-5972-81 B7-H4 Monoclonal Antibody (188), Functional Grade, eBioscience™ 50 µg
192.95
Special offer
Online exclusive
227.00
Save 34.05 (15%)
MA513493 CD155 Monoclonal Antibody (D171) 500 µL
455.00
MA523859 CD153 Monoclonal Antibody (116614) 100 µg
466.00
16-0861-85 CD86 (B7-2) Monoclonal Antibody (PO3.1), Functional Grade, eBioscience™ 500 µg
425.00
MA517853 CD40 Monoclonal Antibody (3/23), Biotin 100 µg
186.00
PA547291 4-1BB Ligand Polyclonal Antibody 100 µg
487.00
PA536393 CD226 Polyclonal Antibody 100 µL
502.00
12-1528-42 CD152 (CTLA-4) Monoclonal Antibody (eBio20A (20A, A3)), PE, eBioscience™ 100 tests
305.15
Special offer
Online exclusive
359.00
Save 53.85 (15%)
11-1379-42 CD137 (4-1BB) Monoclonal Antibody (4B4 (4B4-1)), FITC, eBioscience™ 100 tests
269.45
Special offer
Online exclusive
317.00
Save 47.55 (15%)
16-0960-82 CD96 (TACTILE) Monoclonal Antibody (6A6), Functional Grade, eBioscience™ 100 µg
192.65
Online Exclusive
257.00
Save 64.35 (25%)
46-5901-82 CD137 Ligand (4-1BB Ligand) Monoclonal Antibody (TKS-1), PerCP-eFluor™ 710, eBioscience™ 100 µg
318.75
Special offer
Online exclusive
375.00
Save 56.25 (15%)
MA174439 B7-H4 Monoclonal Antibody (MIH43) 200 µg -
12-5972-82 B7-H4 Monoclonal Antibody (188), PE, eBioscience™ 100 µg
391.00
Special offer
Online exclusive
460.00
Save 69.00 (15%)
12-0861-83 CD86 (B7-2) Monoclonal Antibody (PO3.1), PE, eBioscience™ 200 µg
396.00
16-0281-86 CD28 Monoclonal Antibody (37.51), Functional Grade, eBioscience™ 1 mg
486.65
Online Exclusive
648.00
Save 161.35 (25%)
PA547967 CD137 Polyclonal Antibody 100 µg
487.00
12-0960-80 CD96 (TACTILE) Monoclonal Antibody (6A6), PE, eBioscience™ 25 µg
178.65
Online Exclusive
184.00
Save 5.35 (3%)
14-1341-85 CD134 (OX40) Monoclonal Antibody (OX-86), eBioscience™ 500 µg
340.65
Online Exclusive
350.00
Save 9.35 (3%)
A18356 CD153 Monoclonal Antibody (RM153), PE 50 µg -
17-1551-82 CD155 Monoclonal Antibody (TX56), APC, eBioscience™ 100 µg
349.35
Special offer
Online exclusive
411.00
Save 61.65 (15%)
11-0309-42 CD30 Monoclonal Antibody (Ber-H2), FITC, eBioscience™ 100 tests
224.40
Special offer
Online exclusive
264.00
Save 39.60 (15%)
PA547045 CD153 Polyclonal Antibody 100 µg
483.00
MA180926 CD40 Monoclonal Antibody (LOB7/6) 200 µg
532.00
14-1347-82 CD134 (OX40) Monoclonal Antibody (ACT35 (ACT-35)), eBioscience™ 100 µg
115.65
Online Exclusive
154.00
Save 38.35 (25%)
46-0969-42 CD96 (TACTILE) Monoclonal Antibody (NK92.39), PerCP-eFluor™ 710, eBioscience™ 100 tests
415.65
Special offer
Online exclusive
489.00
Save 73.35 (15%)
MA524315 CD155 Monoclonal Antibody (690912) 100 µg
498.00
14-0801-85 CD80 (B7-1) Monoclonal Antibody (16-10A1), eBioscience™ 500 µg
272.85
Special offer
Online exclusive
321.00
Save 48.15 (15%)
MA517904 CD27 Monoclonal Antibody (LG.3A10), FITC 100 µg
228.00
16-0272-85 CD27 Monoclonal Antibody (LG.3A10), Functional Grade, eBioscience™ 500 µg
422.00
MA517902 CD27 Monoclonal Antibody (LG.3A10) 250 µg
435.00
12-5950-82 CD272 (BTLA) Monoclonal Antibody (6F7), PE, eBioscience™ 100 µg
418.00
MA181186 CD86 Monoclonal Antibody (BU63), FITC 100 µg
467.00
PA547037 CD137 Polyclonal Antibody 100 µg
540.00
MA524281 CD96 Monoclonal Antibody (630612) 100 µg
398.00
MA524280 CD96 Monoclonal Antibody (628211) 100 µg
348.65
Online Exclusive
465.00
Save 116.35 (25%)
46-1550-42 CD155 Monoclonal Antibody (2H7CD155), PerCP-eFluor™ 710, eBioscience™ 100 tests
423.30
Special offer
Online exclusive
498.00
Save 74.70 (15%)
61-1522-82 CD152 (CTLA-4) Monoclonal Antibody (UC10-4B9), PE-eFluor™ 610, eBioscience™ 100 µg
366.35
Special offer
Online exclusive
431.00
Save 64.65 (15%)
MA135890 Nectin 2 Monoclonal Antibody (R2.525) 200 µg
1,096.00
PA547032 VISTA Polyclonal Antibody 100 µg
540.00
MA517990 CD226 Monoclonal Antibody (15F.10E5) 250 µg -
16-0301-85 CD30 Monoclonal Antibody (mCD30.1), Functional Grade, eBioscience™ 500 µg
310.65
Online Exclusive
414.00
Save 103.35 (25%)
11-0862-85 CD86 (B7-2) Monoclonal Antibody (GL1), FITC, eBioscience™ 500 µg
314.50
Special offer
Online exclusive
370.00
Save 55.50 (15%)
17-1128-42 CD112 (Nectin-2) Monoclonal Antibody (R2.477), APC, eBioscience™ 100 tests
346.80
Special offer
Online exclusive
408.00
Save 61.20 (15%)
MA515693 B7-H3 Monoclonal Antibody (6A1) 100 µL
546.00
11-0279-42 CD27 Monoclonal Antibody (O323), FITC, eBioscience™ 100 tests
197.20
Special offer
Online exclusive
232.00
Save 34.80 (15%)
16-1529-82 CD152 (CTLA-4) Monoclonal Antibody (14D3), Functional Grade, eBioscience™ 100 µg
302.60
Special offer
Online exclusive
356.00
Save 53.40 (15%)
16-1541-85 CD154 (CD40 Ligand) Monoclonal Antibody (MR1), Functional Grade, eBioscience™ 500 µg
417.00
50-0709-42 CD70 Monoclonal Antibody (113-16), eFluor™ 660, eBioscience™ 100 tests
356.65
Online Exclusive
475.00
Save 118.35 (25%)
46-2769-42 CD276 (B7-H3) Monoclonal Antibody (7-517), PerCP-eFluor™ 710, eBioscience™ 100 tests
346.80
Special offer
Online exclusive
408.00
Save 61.20 (15%)
16-5949-82 B7-H4 Monoclonal Antibody (H74), Functional Grade, eBioscience™ 100 µg
333.65
Online Exclusive
445.00
Save 111.35 (25%)
MA523591 OX40 Monoclonal Antibody (443318), APC 100 tests
425.00
MA517916 OX40 Monoclonal Antibody (OX-86), FITC 100 µg
215.00
14-1531-85 CD153 Monoclonal Antibody (RM153), eBioscience™ 500 µg
271.15
Special offer
Online exclusive
319.00
Save 47.85 (15%)
MA524284 CD223 Monoclonal Antibody (631501) 100 µg
477.00
12-5970-83 B7-H4 Monoclonal Antibody (Clone 9), PE, eBioscience™ 200 µg
520.00
17-5956-82 CD272 (BTLA) Monoclonal Antibody (8F4), APC, eBioscience™ 100 µg
367.20
Special offer
Online exclusive
432.00
Save 64.80 (15%)
16-1521-85 CD152 (CTLA-4) Monoclonal Antibody (9H10), Functional Grade, eBioscience™ 500 µg
383.00
MA119215 CD80 Monoclonal Antibody (MEM-233) 100 µg
398.00
PA522248 BTLA Polyclonal Antibody 100 µL
494.00
PA547547 CTLA-4 Polyclonal Antibody 100 µg
494.00
12-0301-82 CD30 Monoclonal Antibody (mCD30.1), PE, eBioscience™ 100 µg
210.65
Online Exclusive
281.00
Save 70.35 (25%)
MA174212 BTLA Monoclonal Antibody (MIH26) 100 µg -
PA534516 OX40L Polyclonal Antibody 100 µg
494.00
PA564286 CD96 Polyclonal Antibody 100 µL -
16-5973-81 CD276 (B7-H3) Monoclonal Antibody (M3.2D7), Functional Grade, eBioscience™ 50 µg
169.65
Online Exclusive
226.00
Save 56.35 (25%)
13-5979-80 CD272 (BTLA) Monoclonal Antibody (MIH26), Biotin, eBioscience™ 25 µg
151.65
Online Exclusive
156.00
Save 4.35 (3%)
MA517005 CD28 Monoclonal Antibody (YTH913.12) 500 µg
750.00
16-5950-82 CD272 (BTLA) Monoclonal Antibody (6F7), Functional Grade, eBioscience™ 100 µg
357.00
Special offer
Online exclusive
420.00
Save 63.00 (15%)
PA531111 CD226 Polyclonal Antibody 100 µL
540.00
MA170090 CD80 Monoclonal Antibody (RMMP-1) 200 µg
479.00
16-2231-85 CD223 (LAG-3) Monoclonal Antibody (eBioC9B7W (C9B7W)), Functional Grade, eBioscience™ 500 µg
355.30
Special offer
Online exclusive
418.00
Save 62.70 (15%)
11-0801-86 CD80 (B7-1) Monoclonal Antibody (16-10A1), FITC, eBioscience™ 1 mg
571.20
Special offer
Online exclusive
672.00
Save 100.80 (15%)
14-0862-85 CD86 (B7-2) Monoclonal Antibody (GL1), eBioscience™ 500 µg
231.65
Online Exclusive
308.00
Save 76.35 (25%)
16-0409-85 CD40 Monoclonal Antibody (5C3), Functional Grade, eBioscience™ 500 µg
401.20
Special offer
Online exclusive
472.00
Save 70.80 (15%)
MA110108 CD154 (CD40 Ligand) Monoclonal Antibody (MR-1) 100 µg
307.00
12-1341-83 CD134 (OX40) Monoclonal Antibody (OX-86), PE, eBioscience™ 200 µg
412.00
11-1347-42 CD134 (OX40) Monoclonal Antibody (ACT35 (ACT-35)), FITC, eBioscience™ 100 tests
251.60
Special offer
Online exclusive
296.00
Save 44.40 (15%)
14-0701-85 CD70 Monoclonal Antibody (FR70), eBioscience™ 500 µg
260.65
Online Exclusive
347.00
Save 86.35 (25%)
MA174441 B7-H3 Monoclonal Antibody (MIH42) 200 µg -
HMCD15201 CTLA-4 Monoclonal Antibody (1B8), FITC 500 µg
590.00
PA524552 OX40L Polyclonal Antibody 400 µL
477.00
MA170087 OX40 Monoclonal Antibody (OX86) 250 µg
455.00
MA134061 CD28 Monoclonal Antibody (37.51.1) 200 µg -
MA174440 B7-H4 Monoclonal Antibody (MIH43) 100 µg -
MA516843 BTLA Monoclonal Antibody (MIH26) 200 µg -
17-5949-42 B7-H4 Monoclonal Antibody (H74), APC, eBioscience™ 100 tests
357.00
Special offer
Online exclusive
420.00
Save 63.00 (15%)
MA133895 CD154 (CD40 Ligand) Monoclonal Antibody (39H5) 100 µg -
MA515512 CD80 Monoclonal Antibody (2A2) 100 µL
397.65
Online Exclusive
530.00
Save 132.35 (25%)
16-0271-85 CD27 Monoclonal Antibody (LG.7F9), Functional Grade, eBioscience™ 500 µg
328.10
Special offer
Online exclusive
386.00
Save 57.90 (15%)
16-0402-86 CD40 Monoclonal Antibody (HM40-3), Functional Grade, eBioscience™ 1 mg
637.50
Special offer
Online exclusive
750.00
Save 112.50 (15%)
MA516845 B7-H4 Monoclonal Antibody (MIH43), FITC 100 µg -
14-0869-82 CD86 (B7-2) Monoclonal Antibody (IT2.2), eBioscience™ 100 µg
195.65
Online Exclusive
260.00
Save 64.35 (25%)
700121 TRAF3 Recombinant Rabbit Monoclonal Antibody (12H13L59) 100 µg
528.00
MA513739 CD137 Monoclonal Antibody (BBK-2) 500 µL
370.65
Online Exclusive
494.00
Save 123.35 (25%)
16-5937-85 CD276 (B7-H3) Monoclonal Antibody (MIH35), Functional Grade, eBioscience™ 500 µg
406.30
Special offer
Online exclusive
478.00
Save 71.70 (15%)
14-0281-86 CD28 Monoclonal Antibody (37.51), eBioscience™ 1 mg
476.00
Special offer
Online exclusive
560.00
Save 84.00 (15%)
16-5979-82 CD272 (BTLA) Monoclonal Antibody (MIH26), Functional Grade, eBioscience™ 100 µg
357.00
Special offer
Online exclusive
420.00
Save 63.00 (15%)
12-0869-42 CD86 (B7-2) Monoclonal Antibody (IT2.2), PE, eBioscience™ 100 tests
208.25
Special offer
Online exclusive
245.00
Save 36.75 (15%)
16-1341-85 CD134 (OX40) Monoclonal Antibody (OX-86), Functional Grade, eBioscience™ 500 µg
447.00
25-1529-42 CD152 (CTLA-4) Monoclonal Antibody (14D3), PE-Cyanine7, eBioscience™ 100 tests
447.10
Special offer
Online exclusive
526.00
Save 78.90 (15%)
MA110166 CD28 Monoclonal Antibody (CD28.2) 100 µg
252.00
46-0401-82 CD40 Monoclonal Antibody (1C10), PerCP-eFluor™ 710, eBioscience™ 100 µg
384.20
Special offer
Online exclusive
452.00
Save 67.80 (15%)
PA513483 CD154 (CD40 Ligand) Polyclonal Antibody 400 µL
487.00
PA552493 VISTA Polyclonal Antibody 100 µL
548.00
11-0409-42 CD40 Monoclonal Antibody (5C3), FITC, eBioscience™ 100 tests
214.20
Special offer
Online exclusive
252.00
Save 37.80 (15%)
PA532701 CD70 Polyclonal Antibody 50 µg
498.00
46-5919-82 VISTA Monoclonal Antibody (MH5A), PerCP-eFluor™ 710, eBioscience™ 100 µg
404.60
Special offer
Online exclusive
476.00
Save 71.40 (15%)
MA110172 CD28 Monoclonal Antibody (37.51) 100 µg
217.00
MA110293 CD86 Monoclonal Antibody (BU63) 100 µg
252.00
12-0271-83 CD27 Monoclonal Antibody (LG.7F9), PE, eBioscience™ 200 µg
345.10
Special offer
Online exclusive
406.00
Save 60.90 (15%)
MA110170 CD28 Monoclonal Antibody (CD28.2), PE 100 tests
469.00
Article download
Download a printer-friendly version of this article.
Download now
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
For Research Use Only. Not for use in diagnostic procedures.