Search Thermo Fisher Scientific
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Inorganic Salts | |||
Potassium Chloride (KCl) | 75.0 | 2000.0 | 26.666666 |
Potassium Phosphate monobasic (KH2PO4) | 136.0 | 2000.0 | 14.705882 |
Sodium Chloride (NaCl) | 58.0 | 80000.0 | 1379.3103 |
Sodium Phosphate dibasic (Na2HPO4-7H2O) | 268.0 | 21600.0 | 80.597015 |
1. Dulbecco, R. and Vogt, M., (1954) Plaque formation and isolation of pure lines with Poliomyelitis viruses. J. Exp. Med., 98:167.
14190 - DPBS, no calcium, no magnesium
Cat No:
14190086
,
14190094
,
14190144
,
14190169
,
14190185
,
14190235
,
14190250
,
14190342
,
14190359
,
14190367
21600 - DPBS, powder, no calcium, no magnesium
Cat No:
21600010
,
21600044
,
21600069