CyQUANT® NF Cell Proliferation Assay protocol
Designed for high throughput applications with multi-well plates (96, 384 or 1,536 plate formats), the CyQUANT® NF Cell Proliferation Assay protocol is work-flow friendly. There are no requirements for wash steps or cell lysis. The reagent is added directly to cells after medium removal, incubated for 30 to 60 minutes and read on standard plate readers.
Designed for high throughput applications with multi-well plates (96, 384 or 1,536 plate formats), the CyQUANT® NF Cell Proliferation Assay protocol is work-flow friendly. There are no requirements for wash steps or cell lysis. The reagent is added directly to cells after medium removal, incubated for 30 to 60 minutes and read on standard plate readers.
Related Products
Catalog # Name Size Price (USD) Qty
C35006 CyQUANT™ NF Cell Proliferation Assay, 1,000 assays Each
529.65
Online Exclusive
546.00
Save 16.35 (3%)
Related Images