Nuclear, cytoskeletal, and mitochondrial staining of Muntjac fibroblasts.
Multicolor fluorescence microscopy analysis of Muntjac fibroblasts. The actin cytoskeleton was labeled with biotin phalloidin (Cat. no. B7474) and visualized using Alexa Fluor® 350 streptavidin (Cat. no. S11249). Mitochondria were stained with MitoTracker Red CMXRos (Cat. no. M7512); the nucleus was labeled with an anti-phosphohistone primary antibody and visualized using Alexa Fluor® 500 goat anti–mouse IgG (Cat. no. A31554).
Multicolor fluorescence microscopy analysis of Muntjac fibroblasts. The actin cytoskeleton was labeled with biotin phalloidin (Cat. no. B7474) and visualized using Alexa Fluor® 350 streptavidin (Cat. no. S11249). Mitochondria were stained with MitoTracker Red CMXRos (Cat. no. M7512); the nucleus was labeled with an anti-phosphohistone primary antibody and visualized using Alexa Fluor® 500 goat anti–mouse IgG (Cat. no. A31554).
Related Products
Catalog # Name Size Price (USD) Qty
B7474 Biotin-XX Phalloidin Each
611.65
Online Exclusive
630.00
Save 18.35 (3%)
M7512 MitoTracker™ Red CMXRos - Special Packaging, 20 x 50 μg, 20 x 50 μg Each
506.65
Online Exclusive
522.00
Save 15.35 (3%)
S11249 Streptavidin, Alexa Fluor™ 350 conjugate Each
343.00
Related Images