Knorr Pyrrole Synthesis – a heterocycle formation reaction
In 1886, L. Knorr heated a mixture of alpha-nitroso ethyl acetoacetate and ethyl acetoacetate together in glacial acetic acid in the presence of zinc dust, forming a tetra-substituted pyrrole. Since then, the condensation of an alpha-amino ketone or alpha-amino-beta ketoester with an active methylene compound has been known as the Knorr pyrrole synthesis.
The Neber rearrangement can be used to prepare the precursor alpha-aminoketones from the corresponding oxime by reaction with tosyl chloride.
α-aminoketones are often quite labile and can undergo self-condensation, forming a pyrazine; so, they are often prepared by first nitrosating the ketone, and then reducing it in situ.
An anti-inflammatory analgesic compound, 4,5,8,9-tetrahydro-8-methyl-9-oxothieno[3’3’:5,6]cyclohepta[1,2-b]-pyrrole-7-acetic acid, was synthesized by H.E. Rosenberg and R.W. Ward utilizing the Knorr pyrrole synthesis.
Mechanism of the Knorr pyrrole synthesis
Review available Thermo Scientific products for the Knorr pyrrole synthesis:
Catalog # Name Size Price (USD) Qty
A12544.30 Ethyl acetoacetate, 99+% Each
24.65
Special offer
Online exclusive
Ends: 26-Sep-2025
28.70
Save 4.05 (14%)
A12544.36 Ethyl acetoacetate, 99+% Each
29.65
Special offer
Online exclusive
Ends: 26-Sep-2025
35.10
Save 5.45 (16%)
A12544.0E Ethyl acetoacetate, 99+% Each
84.65
Special offer
Online exclusive
Ends: 26-Sep-2025
98.90
Save 14.25 (14%)
A12544.0C Ethyl acetoacetate, 99+% Each
289.65
Special offer
Online exclusive
Ends: 30-Jun-2025
340.00
Save 50.35 (15%)
L13310.22 Zinc powder, -100 mesh, 97+%, Thermo Scientific Chemicals Each -
L13310.36 Zinc powder, -100 mesh, 97+%, Thermo Scientific Chemicals Each -
A14547.30 p-Toluenesulfonyl chloride, 98% Each
27.65
Special offer
Online exclusive
Ends: 30-Jun-2025
32.40
Save 4.75 (15%)
A14547.36 p-Toluenesulfonyl chloride, 98% Each
48.65
Online Exclusive
53.70
Save 5.05 (9%)
A14547.0B p-Toluenesulfonyl chloride, 98% Each
74.65
Special offer
Online exclusive
Ends: 30-Jun-2025
88.10
Save 13.45 (15%)
A14547.0I p-Toluenesulfonyl chloride, 98% Each
260.65
Special offer
Online exclusive
Ends: 26-Sep-2025
307.00
Save 46.35 (15%)
33381.14 -
33381.22 Each -
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers.  Do you have a favorite reaction?  Let us know!