Overview
The Baeyer-Villiger Oxidation is a prominent process for the oxidation of ketones.
Baeyer-Villiger Oxidation overview
Named Reactions in Organic Synthesis
Baeyer-Villiger Oxidation Mechanism
Watch our video to learn more about the Baeyer-Villiger Oxidation and its mechanisms. 
Video Player is loading.
Current Time 0:00
Duration 2:32
Loaded: 0.00%
Stream Type LIVE
Remaining Time 2:32
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Need reagents for the Baeyer-Villiger Oxidation?
    Chem dex
    If you need further help to plan your synthesis, don’t miss our new app.  Chem dex is a new tool to explore hundreds of chemical reactions, and to have access to product recommendations. Visit: thermofisher.com/chemdex.
    Named Reactions eBook
    Named Reactions in Organic Synthesis eBook
    Download our Named Reactions eBook to learn more about the Baeyer-Villiger Oxidation.
    Access 10 chapters of content about organic chemistry reactions that are named after the chemists that discovered them. This informative eBook includes quizzes so you can test your knowledge!
    History of the Baeyer-Villiger Oxidation
    While exploring the ring cleavage of cyclic ketones in 1899, A. Baeyer and V. Villiger discovered that ketones could be transformed into esters, and cyclic ketones into lactones or hydroxy acids, by using peroxyacids. This reaction became known as the Baeyer-Villiger oxidation. The oxidation of ketones by this method has several benefits:
    • It tolerates the presence of many other functional groups
    • The rearrangement steps retain the existing stereochemistry at the migrating center
    • A wide variety of different peroxyacids can be used as oxidants
    • The oxidation can be performed asymmetrically on racemic or prochiral ketones using enzymes or chiral transition metal catalysts
    • A wide range of oxidizing agents can be used
    • The activity of suitable oxidizing agents can be ranked in the following order: Trifluoroperoxyacetic acid > monopermaleic acid > monoperphthalic acid > 3,5-dinitroperbenzoic acid > p-nitroperbenzoic acid > meta chloroperoxybenzoic acid (mCPBA) > performic acid > perbenzoic acid > peracetic acid > hydrogen peroxide > tert-butyl hydroperoxide (tBuOOH)
    Only a few methods are known for the synthesis of cage-annulated ethers. A.P Marchand and colleagues used the Baeyer-Villiger oxidation for the preparation of novel cage heterocyclic systems, and developed a general process to make cage ethers from cage ketones.
    Other Nucleophilic Substitution Reactions
    Other Nucleophilic Substitution Reactions include: 
    For other types of reactions, visit our Named Reactions page.
    Acroseal Packaging
    Chemical reactions often involve the use of air- and moisture-sensitive solvents, and pyrophoric or hazardous reagents. Our AcroSeal packaging is a packaging solution designed to enable safe handling of these types of materials which are used in a variety of research and development applications, including NMR analysis and studies in drug discovery, agrochemicals, flavors and fragrances, and more. 
    Watch our video for more information. 
    Video Player is loading.
    Current Time 0:00
    Duration 4:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 4:00