Eschenmoser-Claisen Rearrangement
In 1964, the Swiss chemist Albert Eschenmoser reported that allylic or benzylic alcohols heated in the presence of N,N-dimethylacetamide dimethyl acetal in xylenes underwent a form of Claisen rearrangement. This generated a γ,δ-unsaturated amide with a high level of stereospecificity, and today this transformation has become known as the Eschenmoser-Claisen rearrangement.
This rearrangement typically occurs at lower temperatures, between 100-150°C, than other variants such as the Claisen rearrangement itself and is more (E)-selective.
During the first total synthesis of (±)-Stenine by D.J. Hart and colleagues, one of the key steps was an Eschenmoser-Claisen rearrangement that afforded a key amide intermediate in 93% yield.
Mechanism of the Eschenmoser-Claisen rearrangement reaction
Review available Thermo Scientific products for the Eschenmoser-Claisen rearrangement:
Catalog # Name Size Price (USD) Qty
L01642.06 N,N-Dimethylacetamide dimethyl acetal, tech. 90%, stab. Each
55.30
L01642.14 N,N-Dimethylacetamide dimethyl acetal, tech. 90%, stab. Each
145.65
Special offer
Online exclusive
Ends: 30-Jun-2025
171.00
Save 25.35 (15%)
L13317.AP Xylenes, mixed, 97+% Each
31.65
Online Exclusive
35.40
Save 3.75 (11%)
L13317.AU Xylenes, mixed, 97+% Each
39.65
Special offer
Online exclusive
Ends: 26-Sep-2025
46.30
Save 6.65 (14%)
L13317.0F Xylenes, mixed, 97+% Each
57.65
Special offer
Online exclusive
Ends: 26-Sep-2025
67.60
Save 9.95 (15%)
Vote for your favorite named reaction
Learn more about the most popular named reactions in chemistry based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote now